Assay Details
| Gene Symbol: MAN2B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173498.1 | NP_001166969.1 | 7 - 8 | 1100 | 89 | |
| GenBank mRNA | U05572.1 | - | 7 - 8 | 1057 | 89 | |
More Information
Set Membership: |
|

MAN2B1
Genomic Map