Assay Details
| Gene Symbol: MRPL39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017446.3 | NP_059142.2 | 8 - 9 | 934 | 123 | |
| XM_011529651.2 | XP_011527953.1 | 8 - 9 | 1005 | 123 | ||
| GenBank mRNA | AF109357.1 | - | 8 - 9 | 908 | 123 | |
| AF239727.1 | - | 8 - 9 | 920 | 123 | ||
| AK000458.1 | - | 8 - 9 | 925 | 123 | ||
| AK225878.1 | - | 8 - 9 | 925 | 123 | ||
| BC004896.2 | - | 8 - 9 | 907 | 123 | ||
| BC107719.1 | - | 8 - 9 | 930 | 6737868 | 123 | |
| CD368044.1 | - | 2 - 3 | 123 | |||
More Information
Updated Mapping Information
Set Membership: |
|

MRPL39
Genomic Map