| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21003594.hg.1 |
ENST00000339775.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21003594.hg.1 |
ENST00000348354.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21003594.hg.1 |
ENST00000464058.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21003594.hg.1 |
NM_001130914.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21003594.hg.1 |
NM_006806.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21003594.hg.1 |
TC21000299.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21003590.hg.1 |
ENST00000339775.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21003590.hg.1 |
NM_001130914.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21003590.hg.1 |
TC21000299.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
ENST00000339775.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
ENST00000339775.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
ENST00000339775.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
ENST00000339775.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
ENST00000339775.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
ENST00000339775.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
ENST00000339775.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
ENST00000339775.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
ENST00000339775.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
ENST00000348354.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
ENST00000348354.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
ENST00000348354.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
ENST00000348354.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
ENST00000348354.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
ENST00000348354.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
ENST00000348354.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
ENST00000348354.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
ENST00000348354.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
ENST00000457956.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
ENST00000457956.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
ENST00000457956.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
ENST00000457956.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
ENST00000464058.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
ENST00000464058.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
ENST00000464058.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
ENST00000464058.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
ENST00000471860.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
ENST00000471860.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
ENST00000471860.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
ENST00000471860.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
ENST00000496601.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
ENST00000496601.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
ENST00000496601.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
NM_001130914.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
NM_001130914.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
NM_001130914.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
NM_001130914.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
NM_001130914.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
NM_001130914.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
NM_001130914.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
NM_001130914.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
NM_001130914.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
NM_001130914.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
NM_006806.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
NM_006806.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
NM_006806.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
NM_006806.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
NM_006806.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
NM_006806.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
NM_006806.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
NM_006806.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100007663.hg.1 |
NM_006806.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21003591.hg.1 |
ENST00000339775.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21003591.hg.1 |
ENST00000348354.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21003591.hg.1 |
NM_001130914.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21003591.hg.1 |
NM_006806.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21003591.hg.1 |
TC21000299.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR2100151956.hg.1 |
ENST00000339775.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100151956.hg.1 |
ENST00000348354.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100151956.hg.1 |
ENST00000496601.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100151956.hg.1 |
NM_001130914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100151956.hg.1 |
NM_006806.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100151956.hg.1 |
TC2100007663.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100151956.hg.1 |
NM_001130914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100151956.hg.1 |
ENST00000339775.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100151956.hg.1 |
ENST00000348354.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100151956.hg.1 |
ENST00000496601.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100151956.hg.1 |
NM_006806.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100151955.hg.1 |
ENST00000339775.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100151955.hg.1 |
NM_001130914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100151955.hg.1 |
TC2100007663.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100151955.hg.1 |
ENST00000339775.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100151955.hg.1 |
NM_001130914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100151959.hg.1 |
ENST00000339775.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100151959.hg.1 |
ENST00000348354.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100151959.hg.1 |
ENST00000464058.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100151959.hg.1 |
NM_001130914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100151959.hg.1 |
NM_006806.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100151959.hg.1 |
TC2100007663.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100151959.hg.1 |
ENST00000339775.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100151959.hg.1 |
ENST00000348354.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100151959.hg.1 |
ENST00000464058.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100151959.hg.1 |
NM_001130914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100151959.hg.1 |
NM_006806.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
ENST00000339775.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
ENST00000339775.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
ENST00000339775.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
ENST00000339775.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
ENST00000339775.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
ENST00000339775.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
ENST00000339775.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
ENST00000339775.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
ENST00000339775.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
ENST00000348354.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
ENST00000348354.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
ENST00000348354.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
ENST00000348354.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
ENST00000348354.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
ENST00000348354.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
ENST00000348354.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
ENST00000348354.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
ENST00000348354.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
ENST00000457956.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
ENST00000457956.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
ENST00000457956.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
ENST00000457956.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
ENST00000464058.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
ENST00000464058.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
ENST00000464058.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
ENST00000464058.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
ENST00000471860.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
ENST00000471860.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
ENST00000471860.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
ENST00000471860.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
ENST00000496601.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
ENST00000496601.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
ENST00000496601.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
NM_001130914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
NM_001130914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
NM_001130914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
NM_001130914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
NM_001130914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
NM_001130914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
NM_001130914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
NM_001130914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
NM_001130914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
NM_001130914.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
NM_006806.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
NM_006806.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
NM_006806.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
NM_006806.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
NM_006806.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
NM_006806.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
NM_006806.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
NM_006806.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100007663.hg.1 |
NM_006806.4 |
Refseq |