Assay Details
| Gene Symbol: BTG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130914.1 | NP_001124386.1 | 1 - 2 | 250 | 67 | |
| NM_006806.4 | NP_006797.3 | 1 - 2 | 250 | 67 | ||
| GenBank mRNA | AK289408.1 | - | 1 - 2 | 68 | ||
| BC011957.1 | - | 1 - 2 | 144 | 3859781 | 67 | |
| DA381239.1 | - | 1 - 2 | 251 | 67 | ||
| DR155504.1 | - | 1 - 2 | 166 | 67 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC21000299.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK289408.1,BC011957.1,NM_001130914.1,NM_006806.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR21003595.hg.1 | ENST00000339775.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003595.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003595.hg.1 | ENST00000464058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003595.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003595.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003595.hg.1 | TC21000299.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003599.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003599.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003599.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003599.hg.1 | TC21000299.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008372.hg.1 | ENST00000435859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008372.hg.1 | ENST00000435859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003597.hg.1 | ENST00000339775.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003597.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003597.hg.1 | ENST00000464058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003597.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003597.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003597.hg.1 | TC21000299.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000457956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000457956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000457956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000457956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000464058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000464058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000464058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000464058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000471860.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000471860.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000471860.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000471860.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000496601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000496601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | ENST00000496601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007663.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008372.hg.1 | ENST00000435859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008372.hg.1 | ENST00000435859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000457956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000457956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000457956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000457956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000464058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000464058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000464058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000464058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000471860.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000471860.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000471860.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000471860.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000496601.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000496601.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | ENST00000496601.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007663.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100151963.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100151963.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100151963.hg.1 | ENST00000457956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100151963.hg.1 | ENST00000464058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100151963.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100151963.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100151963.hg.1 | TC2100007663.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100151963.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100151963.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100151963.hg.1 | ENST00000457956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100151963.hg.1 | ENST00000464058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100151963.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100151963.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100151965.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100151965.hg.1 | ENST00000457956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100151965.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100151965.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100151965.hg.1 | TC2100007663.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100151965.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100151965.hg.1 | ENST00000457956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100151965.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100151965.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100151960.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100151960.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100151960.hg.1 | ENST00000457956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100151960.hg.1 | ENST00000464058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100151960.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100151960.hg.1 | NM_006806.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100151960.hg.1 | TC2100007663.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100151960.hg.1 | ENST00000339775.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100151960.hg.1 | ENST00000348354.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100151960.hg.1 | ENST00000457956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100151960.hg.1 | ENST00000464058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100151960.hg.1 | NM_001130914.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100151960.hg.1 | NM_006806.4 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 203532 | - | - |

BTG3
Genomic Map