Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 800 | ZNF800 | Anti-sense | Chr. 7 - 127342871 - 127392803 on NCBI Build GRCh38 | ||
| Downstream | uncharacterized LOC100506682 | LOC100506682 | 126.6kb | Sense | Chr. 7 - 127476883 - 127485804 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map