Assay Details
| Gene Symbol: GPR158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020752.2 | NP_065803.2 | 1 - 2 | 960 | 103 | |
| GenBank mRNA | AK095120.1 | - | 1 - 2 | 1258 | 103 | |
| AY528411.1 | - | 1 - 2 | 960 | 103 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000177.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10002052.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002052.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002052.hg.1 | TC10000177.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002053.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002053.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002053.hg.1 | TC10000177.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002054.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002054.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002054.hg.1 | TC10000177.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002057.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002057.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002057.hg.1 | TC10000177.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002055.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002055.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002055.hg.1 | TC10000177.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002059.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002059.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002059.hg.1 | TC10000177.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007061.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007061.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007061.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007061.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007061.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007061.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007061.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007061.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007061.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007061.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007061.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007061.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007061.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007061.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007061.hg.1 | ENST00000482641.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007061.hg.1 | ENST00000482641.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007061.hg.1 | ENST00000482641.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007061.hg.1 | ENST00000490549.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007061.hg.1 | ENST00000490549.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007061.hg.1 | ENST00000490549.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007061.hg.1 | ENST00000490549.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007061.hg.1 | ENST00000490549.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007061.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007061.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007061.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007061.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007061.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007061.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007061.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007061.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007061.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007061.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007061.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007061.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007061.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002051.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002051.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002051.hg.1 | TC10000177.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000148097.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148097.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148097.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148097.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148097.hg.1 | TC1000007061.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000148090.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148090.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148090.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148090.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148090.hg.1 | TC1000007061.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000148093.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148093.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148093.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148093.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148093.hg.1 | TC1000007061.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000148095.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148095.hg.1 | ENST00000482641.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148095.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148095.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148095.hg.1 | ENST00000482641.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148095.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148095.hg.1 | TC1000007061.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000007061.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007061.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007061.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007061.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007061.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007061.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007061.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007061.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007061.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007061.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007061.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007061.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007061.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007061.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007061.hg.1 | ENST00000482641.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007061.hg.1 | ENST00000482641.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007061.hg.1 | ENST00000482641.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007061.hg.1 | ENST00000490549.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007061.hg.1 | ENST00000490549.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007061.hg.1 | ENST00000490549.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007061.hg.1 | ENST00000490549.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007061.hg.1 | ENST00000490549.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007061.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007061.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007061.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007061.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007061.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007061.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007061.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007061.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007061.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007061.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007061.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007061.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007061.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148092.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148092.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148092.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148092.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148092.hg.1 | TC1000007061.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000148089.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148089.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148089.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148089.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148089.hg.1 | TC1000007061.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000148091.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148091.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148091.hg.1 | ENST00000376351.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148091.hg.1 | NM_020752.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148091.hg.1 | TC1000007061.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 220077 | - | - |

GPR158
Genomic Map