Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105372645 | LOC105372645 | -225.2kb | Sense | Chr. 20 - 48471320 - 48477557 on NCBI Build GRCh38 | |
| Downstream | ADP ribosylation factor guanine nucleotide exchange factor 2 | ARFGEF2 |
BIG2, PVNH2, dJ1164I10.1 |
217.1kb | Sense | Chr. 20 - 48921721 - 49036693 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map