Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017958.hg.1 |
ENST00000299641.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017958.hg.1 |
ENST00000309979.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017958.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017958.hg.1 |
TC10001419.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000398701.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000398701.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000398701.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000398701.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000429742.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000429742.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000429742.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000429742.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000463410.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000463410.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000463410.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000465929.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000465929.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017959.hg.1 |
ENST00000299641.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017959.hg.1 |
ENST00000309979.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017959.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017959.hg.1 |
TC10001419.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000398701.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000398701.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000398701.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000398701.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000429742.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000429742.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000429742.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000429742.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000463410.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000463410.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000463410.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000465929.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000465929.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000187413.hg.1 |
ENST00000299641.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000187413.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000187413.hg.1 |
ENST00000429742.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000187413.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000187413.hg.1 |
ENST00000299641.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000187413.hg.1 |
ENST00000309979.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000187413.hg.1 |
ENST00000429742.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000187413.hg.1 |
NM_003635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000187413.hg.1 |
TC1000012570.hg.1 |
Clariom D TC |