| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000398701.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000398701.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000398701.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000398701.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000429742.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000429742.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000429742.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000429742.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000463410.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000463410.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000463410.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000465929.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
ENST00000465929.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10017956.hg.1 |
ENST00000299641.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10017956.hg.1 |
ENST00000309979.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10017956.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10017956.hg.1 |
TC10001419.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1000187412.hg.1 |
ENST00000299641.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000187412.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000187412.hg.1 |
ENST00000429742.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000187412.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000187412.hg.1 |
ENST00000299641.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000187412.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000187412.hg.1 |
ENST00000429742.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000187412.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000187412.hg.1 |
TC1000012570.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000299641.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000309979.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000398701.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000398701.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000398701.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000398701.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000429742.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000429742.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000429742.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000429742.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000463410.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000463410.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000463410.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000465929.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
ENST00000465929.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012570.hg.1 |
NM_003635.3 |
Refseq |