Assay Details
| Gene Symbol: SMARCA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003601.3 | NP_003592.3 | 16 - 17 | 2635 | 131 | |
| GenBank mRNA | AB010882.1 | - | 16 - 17 | 2375 | 131 | |
| AK302841.1 | - | 17 - 18 | 2270 | 131 | ||
| AK316444.1 | - | 17 - 18 | 2437 | 131 | ||
| BC023144.1 | - | 16 - 17 | 2386 | 4340921 | 131 | |
More Information
Set Membership: |
|

SMARCA5
Genomic Map