Assay Details
| Gene Symbol: OGFRL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001324266.1 | NP_001311195.1 | 3 - 4 | 900 | 160 | |
| XM_011536117.2 | XP_011534419.1 | 3 - 4 | 833 | 160 | ||
| XM_011536118.2 | XP_011534420.1 | 3 - 4 | 682 | 160 | ||
| XM_017011303.1 | XP_016866792.1 | 3 - 4 | 462 | 160 | ||
| XM_017011305.1 | XP_016866794.1 | 3 - 4 | 355 | 160 | ||
| XM_017011306.1 | XP_016866795.1 | 3 - 4 | 1385 | 160 | ||
| GenBank mRNA | BX420584.2 | - | 3 - 4 | 566 | 160 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011536117.2, XM_011536118.2, XM_017011306.1 transcripts.
Set Membership: |
|

OGFRL1
Genomic Map