Assay Details
| Gene Symbol: HPCA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017001118.1 | XP_016856607.1 | 1 | 413 | 66 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the XM_017001118.1 transcripts.
This assay is in the 5'UTR region of the XM_017001118.1 transcripts.
Set Membership: |
|

HPCA
Genomic Map