Assay Details
| Gene Symbol: SLC5A12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178498.3 | NP_848593.2 | 13 - 14 | 1891 | 156 | |
| XM_006718155.3 | XP_006718218.1 | 13 - 14 | 1794 | 156 | ||
| XM_011519920.2 | XP_011518222.1 | 14 - 15 | 1371 | 156 | ||
| XM_017017244.1 | XP_016872733.1 | 14 - 15 | 1817 | 156 | ||
| GenBank mRNA | AY299482.1 | - | 13 - 14 | 1600 | 156 | |
| BC041454.1 | - | 13 - 14 | 1364 | 156 | ||
| CR747563.1 | - | 2 - 3 | 276 | 156 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001506.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11018837.hg.1 | ENST00000396005.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11018837.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11018837.hg.1 | TC11001506.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11018836.hg.1 | ENST00000396005.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11018836.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11018836.hg.1 | TC11001506.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100178760.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100178760.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100178760.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100178760.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100178760.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100178760.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100178760.hg.1 | TC1100010393.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100178759.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100178759.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100178759.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100178759.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100178759.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100178759.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100178759.hg.1 | TC1100010393.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000280467.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000396005.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000527405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000528822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | ENST00000533617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010393.hg.1 | NM_178498.3 | Refseq |

SLC5A12
Genomic Map