| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01023142.hg.1 |
ENST00000367749.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01023142.hg.1 |
NM_002022.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01023142.hg.1 |
TC01001490.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
ENST00000367749.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
ENST00000367749.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
ENST00000367749.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
ENST00000367749.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
ENST00000367749.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
ENST00000367749.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
ENST00000367749.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
ENST00000367749.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
ENST00000367749.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
ENST00000367749.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
ENST00000367749.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
ENST00000367749.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
ENST00000367749.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
ENST00000367749.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
ENST00000367749.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
ENST00000462992.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
ENST00000462992.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
ENST00000462992.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
ENST00000462992.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
ENST00000462992.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
ENST00000475780.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
ENST00000475780.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
ENST00000475780.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
ENST00000475780.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
ENST00000475780.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
ENST00000475780.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
ENST00000480136.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
ENST00000480136.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
ENST00000480136.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
ENST00000480136.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
ENST00000497228.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
ENST00000497228.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
ENST00000497228.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
ENST00000497228.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
ENST00000497228.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
NM_002022.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
NM_002022.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
NM_002022.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
NM_002022.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
NM_002022.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
NM_002022.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
NM_002022.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
NM_002022.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
NM_002022.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
NM_002022.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
NM_002022.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
NM_002022.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
NM_002022.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
NM_002022.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
NM_002022.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
NM_002022.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010586.hg.1 |
NM_002022.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100177961.hg.1 |
ENST00000367749.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100177961.hg.1 |
ENST00000480136.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100177961.hg.1 |
NM_002022.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100177961.hg.1 |
ENST00000367749.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100177961.hg.1 |
ENST00000480136.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100177961.hg.1 |
NM_002022.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100177961.hg.1 |
TC0100010586.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
ENST00000367749.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
ENST00000367749.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
ENST00000367749.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
ENST00000367749.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
ENST00000367749.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
ENST00000367749.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
ENST00000367749.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
ENST00000367749.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
ENST00000367749.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
ENST00000367749.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
ENST00000367749.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
ENST00000367749.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
ENST00000367749.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
ENST00000367749.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
ENST00000367749.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
ENST00000462992.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
ENST00000462992.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
ENST00000462992.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
ENST00000462992.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
ENST00000462992.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
ENST00000475780.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
ENST00000475780.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
ENST00000475780.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
ENST00000475780.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
ENST00000475780.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
ENST00000475780.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
ENST00000480136.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
ENST00000480136.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
ENST00000480136.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
ENST00000480136.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
ENST00000497228.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
ENST00000497228.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
ENST00000497228.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
ENST00000497228.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
ENST00000497228.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
NM_002022.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
NM_002022.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
NM_002022.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
NM_002022.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
NM_002022.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
NM_002022.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
NM_002022.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
NM_002022.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
NM_002022.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
NM_002022.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
NM_002022.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
NM_002022.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
NM_002022.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
NM_002022.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
NM_002022.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
NM_002022.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010586.hg.1 |
NM_002022.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100177960.hg.1 |
ENST00000367749.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100177960.hg.1 |
NM_002022.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100177960.hg.1 |
ENST00000367749.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100177960.hg.1 |
NM_002022.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100177960.hg.1 |
TC0100010586.hg.1 |
Clariom D TC |