Assay Details
| Gene Symbol: PPP1R36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172365.2 | NP_758953.1 | 2 - 3 | 229 | 107 | |
| XM_011536468.2 | XP_011534770.1 | 2 - 3 | 197 | 107 | ||
| GenBank mRNA | AK058044.1 | - | 2 - 3 | 229 | 107 | |
| BC068989.1 | - | 2 - 3 | 194 | 30389944 | 107 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000392.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR14004472.hg.1 | ENST00000298705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004472.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004472.hg.1 | TC14000392.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004477.hg.1 | ENST00000298705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004477.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004477.hg.1 | TC14000392.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000554400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000554400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000554400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000554400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000554400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000554400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000555645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000555645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000555645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000555645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000555645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000555645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000556023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000556023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000556023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000556023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004475.hg.1 | ENST00000298705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004475.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004475.hg.1 | TC14000392.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004474.hg.1 | ENST00000298705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004474.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004474.hg.1 | TC14000392.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1400152003.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152003.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152003.hg.1 | ENST00000555645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152003.hg.1 | ENST00000557202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152003.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400152003.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152003.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152003.hg.1 | ENST00000555645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152003.hg.1 | ENST00000557202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152003.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400152003.hg.1 | TC1400007444.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400152005.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152005.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152005.hg.1 | ENST00000555645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152005.hg.1 | ENST00000557202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152005.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400152005.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152005.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152005.hg.1 | ENST00000555645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152005.hg.1 | ENST00000557202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152005.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400152005.hg.1 | TC1400007444.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400152001.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152001.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152001.hg.1 | ENST00000555645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152001.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400152001.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152001.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152001.hg.1 | ENST00000555645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152001.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400152001.hg.1 | TC1400007444.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400152000.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152000.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152000.hg.1 | ENST00000555645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152000.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400152000.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152000.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152000.hg.1 | ENST00000555645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152000.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400152000.hg.1 | TC1400007444.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000298705.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000467261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000554400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000554400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000554400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000554400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000554400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000554400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000555645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000555645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000555645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000555645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000555645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000555645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000556023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000556023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000556023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000556023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | ENST00000557202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | NM_172365.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007444.hg.1 | NM_172365.2 | Refseq |

PPP1R36
Genomic Map