Assay Details
| Gene Symbol: SCAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012235.3 | NP_036367.2 | 16 - 17 | 2712 | 58 | |
| XM_005264968.1 | XP_005265025.1 | 13 - 14 | 2381 | 58 | ||
| XM_005264971.1 | XP_005265028.1 | 10 - 11 | 1792 | 58 | ||
| XM_011533501.1 | XP_011531803.1 | 16 - 17 | 2739 | 58 | ||
| XM_011533502.2 | XP_011531804.1 | 8 - 9 | 1388 | 58 | ||
| XM_017005918.1 | XP_016861407.1 | 16 - 17 | 2726 | 58 | ||
| XM_017005919.1 | XP_016861408.1 | 16 - 17 | 2659 | 58 | ||
| GenBank mRNA | BC027207.2 | - | 11 - 12 | 1405 | 58 | |
| D83782.2 | - | 16 - 17 | 2698 | 58 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001367.hg.1
Set Membership: |
|

SCAP
Genomic Map