| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014281.hg.1 |
ENST00000605528.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014281.hg.1 |
ENST00000605528.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014281.hg.1 |
ENST00000605528.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014281.hg.1 |
ENST00000605528.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014281.hg.1 |
ENST00000605528.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014281.hg.1 |
ENST00000605528.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014281.hg.1 |
ENST00000605528.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014281.hg.1 |
ENST00000605528.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014281.hg.1 |
NM_001202470.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014281.hg.1 |
NM_001202470.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014281.hg.1 |
NM_001202470.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014281.hg.1 |
NM_001202470.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014281.hg.1 |
NM_001202470.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014281.hg.1 |
NM_001202470.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014281.hg.1 |
NM_001202470.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014281.hg.1 |
NM_001202470.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014281.hg.1 |
NM_001202470.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014281.hg.1 |
NM_001202470.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014283.hg.1 |
ENST00000402419.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014283.hg.1 |
ENST00000402419.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014283.hg.1 |
ENST00000607016.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014283.hg.1 |
ENST00000607016.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014283.hg.1 |
ENST00000607016.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014283.hg.1 |
ENST00000607016.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014283.hg.1 |
ENST00000607016.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014283.hg.1 |
ENST00000607016.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014283.hg.1 |
ENST00000607016.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014283.hg.1 |
ENST00000607016.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014283.hg.1 |
ENST00000607016.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014283.hg.1 |
ENST00000607016.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014283.hg.1 |
ENST00000607016.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014283.hg.1 |
ENST00000607016.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014283.hg.1 |
ENST00000607016.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014283.hg.1 |
ENST00000607016.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014283.hg.1 |
ENST00000607016.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014283.hg.1 |
ENST00000607016.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014283.hg.1 |
NM_006703.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014283.hg.1 |
NM_006703.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014283.hg.1 |
NM_006703.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014283.hg.1 |
NM_006703.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014283.hg.1 |
NM_006703.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014283.hg.1 |
NM_006703.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014283.hg.1 |
NM_006703.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014283.hg.1 |
NM_006703.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014283.hg.1 |
NM_006703.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019563.hg.1 |
ENST00000358797.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019563.hg.1 |
NM_006703.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019563.hg.1 |
TC06004129.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0600014283.hg.1 |
ENST00000402419.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014283.hg.1 |
ENST00000402419.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014283.hg.1 |
ENST00000607016.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014283.hg.1 |
ENST00000607016.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014283.hg.1 |
ENST00000607016.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014283.hg.1 |
ENST00000607016.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014283.hg.1 |
ENST00000607016.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014283.hg.1 |
ENST00000607016.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014283.hg.1 |
ENST00000607016.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014283.hg.1 |
ENST00000607016.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014283.hg.1 |
ENST00000607016.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014283.hg.1 |
ENST00000607016.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014283.hg.1 |
ENST00000607016.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014283.hg.1 |
ENST00000607016.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014283.hg.1 |
ENST00000607016.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014283.hg.1 |
ENST00000607016.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014283.hg.1 |
ENST00000607016.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014283.hg.1 |
ENST00000607016.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014283.hg.1 |
NM_006703.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014283.hg.1 |
NM_006703.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014283.hg.1 |
NM_006703.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014283.hg.1 |
NM_006703.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014283.hg.1 |
NM_006703.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014283.hg.1 |
NM_006703.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014283.hg.1 |
NM_006703.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014283.hg.1 |
NM_006703.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014283.hg.1 |
NM_006703.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600201611.hg.1 |
ENST00000605528.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600201611.hg.1 |
NM_001202470.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600201611.hg.1 |
ENST00000605528.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600201611.hg.1 |
NM_001202470.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600201611.hg.1 |
TC0600014281.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600201659.hg.1 |
ENST00000607016.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600201659.hg.1 |
NM_006703.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600201659.hg.1 |
ENST00000607016.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600201659.hg.1 |
NM_006703.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600201659.hg.1 |
TC0600014283.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600014281.hg.1 |
ENST00000605528.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014281.hg.1 |
ENST00000605528.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014281.hg.1 |
ENST00000605528.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014281.hg.1 |
ENST00000605528.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014281.hg.1 |
ENST00000605528.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014281.hg.1 |
ENST00000605528.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014281.hg.1 |
ENST00000605528.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014281.hg.1 |
ENST00000605528.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014281.hg.1 |
NM_001202470.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014281.hg.1 |
NM_001202470.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014281.hg.1 |
NM_001202470.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014281.hg.1 |
NM_001202470.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014281.hg.1 |
NM_001202470.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014281.hg.1 |
NM_001202470.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014281.hg.1 |
NM_001202470.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014281.hg.1 |
NM_001202470.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014281.hg.1 |
NM_001202470.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014281.hg.1 |
NM_001202470.2 |
Refseq |