Assay Details
| Gene Symbol: PSME4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014614.2 | NP_055429.2 | 14 - 15 | 1869 | 98 | |
| XM_006711969.1 | XP_006712032.1 | 12 - 13 | 1385 | 98 | ||
| XM_011532705.2 | XP_011531007.1 | 15 - 16 | 1893 | 98 | ||
| XM_011532706.2 | XP_011531008.1 | 15 - 16 | 1893 | 98 | ||
| XM_011532707.2 | XP_011531009.1 | 14 - 15 | 1827 | 98 | ||
| XM_011532709.2 | XP_011531011.1 | 15 - 16 | 1893 | 98 | ||
| GenBank mRNA | AB621805.1 | - | 13 - 14 | 1986 | 98 | |
| AY894756.1 | - | 14 - 15 | 1813 | 98 | ||
| BC112169.1 | - | 13 - 14 | 1505 | 8327637 | 98 | |
| BC113668.1 | - | 13 - 14 | 1505 | 8322720 | 98 | |
| BC143739.1 | - | 13 - 14 | 1505 | 98 | ||
| D38521.1 | - | 14 - 15 | 1679 | 98 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001848.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_006711969.1 transcripts.
Set Membership: |
|

PSME4
Genomic Map