Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029523.hg.1 |
TC02001848.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029523.hg.1 |
ENST00000404125.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029523.hg.1 |
ENST00000421748.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02029523.hg.1 |
NM_014614.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200191915.hg.1 |
ENST00000389993.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191915.hg.1 |
ENST00000404125.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191915.hg.1 |
NM_014614.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200191915.hg.1 |
ENST00000389993.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191915.hg.1 |
ENST00000404125.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200191915.hg.1 |
NM_014614.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200191915.hg.1 |
TC0200012608.hg.1 |
Clariom D TC |