| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02029519.hg.1 |
ENST00000404125.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02029519.hg.1 |
ENST00000421748.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02029519.hg.1 |
NM_014614.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02029519.hg.1 |
TC02001848.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0200191911.hg.1 |
ENST00000389993.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200191911.hg.1 |
ENST00000404125.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200191911.hg.1 |
NM_014614.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200191911.hg.1 |
ENST00000389993.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200191911.hg.1 |
ENST00000404125.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200191911.hg.1 |
NM_014614.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200191911.hg.1 |
TC0200012608.hg.1 |
Clariom D TC |