| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03019694.hg.1 |
ENST00000360413.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03019694.hg.1 |
ENST00000383836.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03019694.hg.1 |
ENST00000433332.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03019694.hg.1 |
ENST00000544908.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03019694.hg.1 |
NM_001033117.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03019694.hg.1 |
NM_014850.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03019694.hg.1 |
TC03003372.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0300206428.hg.1 |
ENST00000360413.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206428.hg.1 |
ENST00000383836.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206428.hg.1 |
ENST00000433332.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206428.hg.1 |
ENST00000475560.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206428.hg.1 |
ENST00000618999.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206428.hg.1 |
NM_001033117.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300206428.hg.1 |
NM_014850.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300206428.hg.1 |
ENST00000360413.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206428.hg.1 |
ENST00000383836.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206428.hg.1 |
ENST00000433332.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206428.hg.1 |
ENST00000475560.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206428.hg.1 |
ENST00000618999.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300206428.hg.1 |
NM_001033117.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300206428.hg.1 |
NM_014850.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300206428.hg.1 |
TC0300013932.hg.1 |
Clariom D TC |