Assay Details
| Gene Symbol: EPB41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004437.3 | NP_004428.1 | 14 - 15 | 2092 | 134 | |
| XM_005245764.1 | XP_005245821.1 | 13 - 14 | 2104 | 134 | ||
| XM_011540960.1 | XP_011539262.1 | 13 - 14 | 2104 | 134 | ||
| XM_017000594.1 | XP_016856083.1 | 13 - 14 | 2000 | 134 | ||
| GenBank mRNA | BC096105.1 | - | 12 - 13 | 1636 | 40001729 | 134 |
| BC114450.1 | - | 12 - 13 | 1636 | 134 | ||
| M14993.1 | - | 14 - 15 | 2073 | 134 | ||
More Information
Set Membership: |
|

EPB41
Genomic Map