| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01055356.hg.1 |
ENST00000367595.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01055356.hg.1 |
ENST00000475338.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01055356.hg.1 |
NM_032360.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01055356.hg.1 |
TC01006385.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018540.hg.1 |
ENST00000367595.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018540.hg.1 |
ENST00000367595.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018540.hg.1 |
ENST00000367595.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018540.hg.1 |
ENST00000367595.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018540.hg.1 |
ENST00000367595.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018540.hg.1 |
ENST00000367595.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018540.hg.1 |
ENST00000367595.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018540.hg.1 |
ENST00000367595.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018540.hg.1 |
ENST00000367595.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018540.hg.1 |
ENST00000367595.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018540.hg.1 |
ENST00000367595.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018540.hg.1 |
ENST00000367595.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018540.hg.1 |
ENST00000367595.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018540.hg.1 |
ENST00000475338.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018540.hg.1 |
ENST00000475338.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018540.hg.1 |
ENST00000475338.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018540.hg.1 |
ENST00000475338.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018540.hg.1 |
ENST00000475338.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018540.hg.1 |
ENST00000475338.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018540.hg.1 |
ENST00000475338.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018540.hg.1 |
NM_032360.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018540.hg.1 |
NM_032360.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018540.hg.1 |
NM_032360.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018540.hg.1 |
NM_032360.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018540.hg.1 |
NM_032360.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018540.hg.1 |
NM_032360.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018540.hg.1 |
NM_032360.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018540.hg.1 |
NM_032360.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018540.hg.1 |
NM_032360.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018540.hg.1 |
NM_032360.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018540.hg.1 |
NM_032360.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018540.hg.1 |
NM_032360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100244250.hg.1 |
ENST00000367595.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100244250.hg.1 |
ENST00000475338.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100244250.hg.1 |
NM_032360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100244250.hg.1 |
ENST00000367595.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100244250.hg.1 |
ENST00000475338.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100244250.hg.1 |
NM_032360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100244250.hg.1 |
TC0100018540.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100018540.hg.1 |
ENST00000367595.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018540.hg.1 |
ENST00000367595.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018540.hg.1 |
ENST00000367595.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018540.hg.1 |
ENST00000367595.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018540.hg.1 |
ENST00000367595.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018540.hg.1 |
ENST00000367595.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018540.hg.1 |
ENST00000367595.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018540.hg.1 |
ENST00000367595.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018540.hg.1 |
ENST00000367595.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018540.hg.1 |
ENST00000367595.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018540.hg.1 |
ENST00000367595.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018540.hg.1 |
ENST00000367595.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018540.hg.1 |
ENST00000367595.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018540.hg.1 |
ENST00000475338.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018540.hg.1 |
ENST00000475338.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018540.hg.1 |
ENST00000475338.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018540.hg.1 |
ENST00000475338.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018540.hg.1 |
ENST00000475338.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018540.hg.1 |
ENST00000475338.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018540.hg.1 |
ENST00000475338.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018540.hg.1 |
NM_032360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018540.hg.1 |
NM_032360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018540.hg.1 |
NM_032360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018540.hg.1 |
NM_032360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018540.hg.1 |
NM_032360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018540.hg.1 |
NM_032360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018540.hg.1 |
NM_032360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018540.hg.1 |
NM_032360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018540.hg.1 |
NM_032360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018540.hg.1 |
NM_032360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018540.hg.1 |
NM_032360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018540.hg.1 |
NM_032360.3 |
Refseq |