Assay Details
| Gene Symbol: LINC00479 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027272.1 | 3 - 4 | 312 | 130 | ||
| GenBank mRNA | AK057397.1 | - | 3 - 4 | 312 | 130 | |
| BC128198.1 | - | 2 - 3 | 252 | 130 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 111 | LINC00111 |
C21orf21, NCRNA00111 |
-16.7kb | Anti-sense | Chr. 21 - 41679302 - 41697336 on NCBI Build GRCh38 |
| Downstream | long intergenic non-protein coding RNA 112 | LINC00112 |
C21orf22, NCRNA00112 |
2.1kb | Anti-sense | Chr. 21 - 41716436 - 41717582 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC21000475.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR21005706.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21005706.hg.1 | NR_027272.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21005706.hg.1 | TC21000475.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000457881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000457881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000457881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000457881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000457881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000457881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000457881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000457881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000457881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000457881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000457881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000457881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000586552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000586552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000586552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000586552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000589300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000589300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000589300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000589300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000589300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000589300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000589300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | ENST00000589300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | NR_027272.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | NR_027272.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | NR_027272.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | NR_027272.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | NR_027272.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | NR_027272.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | NR_027272.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | NR_027272.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | NR_027272.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | NR_027272.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | NR_027272.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | NR_027272.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | NR_027272.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | NR_027272.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008218.hg.1 | NR_027272.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100155639.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100155639.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100155639.hg.1 | ENST00000589300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100155639.hg.1 | NR_027272.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100155639.hg.1 | TC2100008218.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100155639.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100155639.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100155639.hg.1 | ENST00000589300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100155639.hg.1 | NR_027272.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000412102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000457881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000457881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000457881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000457881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000457881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000457881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000457881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000457881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000457881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000457881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000457881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000457881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000586552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000586552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000586552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000586552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000589300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000589300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000589300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000589300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000589300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000589300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000589300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | ENST00000589300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | NR_027272.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | NR_027272.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | NR_027272.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | NR_027272.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | NR_027272.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | NR_027272.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | NR_027272.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | NR_027272.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | NR_027272.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | NR_027272.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | NR_027272.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | NR_027272.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | NR_027272.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | NR_027272.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008218.hg.1 | NR_027272.1 | Refseq |

LINC00479
Genomic Map