Assay Details
| Gene Symbol: ST14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021978.3 | NP_068813.1 | 11 - 12 | 1544 | 70 | |
| GenBank mRNA | AB030036.1 | - | 11 - 12 | 1366 | 70 | |
| AF057145.1 | - | 11 - 12 | 1373 | 70 | ||
| AF118224.2 | - | 11 - 12 | 1386 | 70 | ||
| AF133086.1 | - | 11 - 12 | 1387 | 70 | ||
| AK314173.1 | - | 11 - 12 | 1527 | 70 | ||
| BC005826.2 | - | 4 - 5 | 364 | 70 | ||
| BC018146.1 | - | 1 - 2 | 53 | 70 | ||
| BC030532.1 | - | 11 - 12 | 1468 | 5213189 | 70 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001195.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000524718.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000524718.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000524718.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000524718.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000524718.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000524718.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000524718.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000524718.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000524718.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000530376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000530376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000530376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000530376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000530376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000530376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000530532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000530532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000530532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000530532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000530532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000530532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000530532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11015046.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11015046.hg.1 | ENST00000525779.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11015046.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11015046.hg.1 | TC11001195.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11015045.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11015045.hg.1 | ENST00000525779.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11015045.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11015045.hg.1 | TC11001195.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100171622.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100171622.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100171622.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100171622.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100171622.hg.1 | TC1100009524.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000524718.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000524718.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000524718.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000524718.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000524718.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000524718.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000524718.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000524718.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000524718.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000530376.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000530376.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000530376.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000530376.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000530376.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000530376.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000530532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000530532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000530532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000530532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000530532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000530532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000530532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100171623.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100171623.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100171623.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100171623.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100171623.hg.1 | TC1100009524.hg.1 | Clariom D TC |

ST14
Genomic Map