| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08016926.hg.1 |
ENST00000276590.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08016926.hg.1 |
ENST00000522447.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08016926.hg.1 |
NM_016027.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08016926.hg.1 |
TC08001313.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
ENST00000276590.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
ENST00000276590.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
ENST00000276590.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
ENST00000276590.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
ENST00000276590.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
ENST00000276590.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
ENST00000276590.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
ENST00000276590.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
ENST00000276590.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
ENST00000276590.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
ENST00000276590.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
ENST00000276590.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
ENST00000517601.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
ENST00000517601.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
ENST00000517601.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
ENST00000522447.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
ENST00000522447.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
ENST00000522447.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
ENST00000522447.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
ENST00000522447.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
ENST00000522447.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
ENST00000522447.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
ENST00000522447.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
ENST00000522447.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
ENST00000522447.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
ENST00000522447.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
ENST00000522558.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
ENST00000522558.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
ENST00000522558.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
NM_016027.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
NM_016027.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
NM_016027.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
NM_016027.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
NM_016027.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
NM_016027.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
NM_016027.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
NM_016027.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
NM_016027.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
NM_016027.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
NM_016027.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
NM_016027.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010766.hg.1 |
NM_016027.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08016927.hg.1 |
ENST00000276590.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08016927.hg.1 |
ENST00000517601.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08016927.hg.1 |
ENST00000522447.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08016927.hg.1 |
NM_016027.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08016927.hg.1 |
TC08001313.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0800173095.hg.1 |
ENST00000276590.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173095.hg.1 |
ENST00000517601.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173095.hg.1 |
ENST00000522447.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173095.hg.1 |
ENST00000522558.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173095.hg.1 |
NM_016027.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800173095.hg.1 |
ENST00000276590.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173095.hg.1 |
ENST00000517601.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173095.hg.1 |
ENST00000522447.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173095.hg.1 |
ENST00000522558.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173095.hg.1 |
NM_016027.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800173095.hg.1 |
TC0800010766.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800173094.hg.1 |
ENST00000276590.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173094.hg.1 |
ENST00000522447.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173094.hg.1 |
ENST00000522558.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173094.hg.1 |
NM_016027.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800173094.hg.1 |
ENST00000276590.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173094.hg.1 |
ENST00000522447.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173094.hg.1 |
ENST00000522558.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173094.hg.1 |
NM_016027.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800173094.hg.1 |
TC0800010766.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
ENST00000276590.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
ENST00000276590.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
ENST00000276590.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
ENST00000276590.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
ENST00000276590.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
ENST00000276590.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
ENST00000276590.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
ENST00000276590.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
ENST00000276590.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
ENST00000276590.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
ENST00000276590.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
ENST00000276590.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
ENST00000517601.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
ENST00000517601.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
ENST00000517601.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
ENST00000522447.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
ENST00000522447.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
ENST00000522447.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
ENST00000522447.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
ENST00000522447.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
ENST00000522447.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
ENST00000522447.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
ENST00000522447.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
ENST00000522447.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
ENST00000522447.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
ENST00000522447.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
ENST00000522558.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
ENST00000522558.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
ENST00000522558.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
NM_016027.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
NM_016027.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
NM_016027.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
NM_016027.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
NM_016027.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
NM_016027.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
NM_016027.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
NM_016027.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
NM_016027.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
NM_016027.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
NM_016027.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
NM_016027.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010766.hg.1 |
NM_016027.2 |
Refseq |