Assay Details
| Gene Symbol: APOBEC3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270411.1 | NP_001257340.1 | 2 - 3 | 82 | ||
| NM_004900.4 | NP_004891.4 | 2 - 3 | 82 | |||
| GenBank mRNA | AK024854.1 | - | 2 - 3 | 82 | ||
| AU098564.1 | - | 2 - 3 | 82 | |||
| AY743217.1 | - | 2 - 3 | 82 | |||
| BC053859.1 | - | 2 - 3 | 254 | 6174285 | 82 | |
| CT841510.1 | - | 2 - 3 | 198 | 82 | ||
| KF651167.1 | - | 2 - 3 | 173 | 82 | ||
| U61084.1 | - | 2 - 3 | 202 | 98 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22001425.hg.1
Additional Information:
This assay is in the 5'UTR region of the U61084.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22005761.hg.1 | ENST00000333467.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005761.hg.1 | ENST00000402182.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005761.hg.1 | ENST00000407298.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005761.hg.1 | NM_004900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005761.hg.1 | TC22001425.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005763.hg.1 | ENST00000333467.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005763.hg.1 | ENST00000402182.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005763.hg.1 | ENST00000407298.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005763.hg.1 | NM_004900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005763.hg.1 | TC22001425.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005764.hg.1 | ENST00000333467.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005764.hg.1 | ENST00000402182.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005764.hg.1 | ENST00000407298.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005764.hg.1 | NM_004900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005764.hg.1 | TC22001425.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000333467.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000333467.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000333467.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000333467.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000333467.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000333467.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000333467.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000333467.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000333467.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000333467.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000333467.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000333467.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000335760.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000335760.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000335760.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000335760.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000335760.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000335760.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000335760.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000335760.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000335760.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000402182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000402182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000402182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000402182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000402182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000402182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000402182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000402182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000402182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000402182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000402182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000402182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000402182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000407298.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000407298.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000407298.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000407298.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000407298.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000407298.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000407298.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000407298.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000407298.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000407298.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | ENST00000407298.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | NM_001270411.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | NM_001270411.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | NM_001270411.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | NM_001270411.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | NM_001270411.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | NM_001270411.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | NM_001270411.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | NM_001270411.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | NM_001270411.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | NM_001270411.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | NM_001270411.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | NM_004900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | NM_004900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | NM_004900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | NM_004900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | NM_004900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | NM_004900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | NM_004900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | NM_004900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | NM_004900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | NM_004900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | NM_004900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009270.hg.1 | NM_004900.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200168861.hg.1 | ENST00000333467.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168861.hg.1 | ENST00000335760.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168861.hg.1 | ENST00000402182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168861.hg.1 | ENST00000407298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168861.hg.1 | NM_001270411.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200168861.hg.1 | NM_004900.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200168861.hg.1 | TC2200009270.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200168861.hg.1 | ENST00000333467.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168861.hg.1 | ENST00000335760.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168861.hg.1 | ENST00000402182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168861.hg.1 | ENST00000407298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168861.hg.1 | NM_001270411.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200168861.hg.1 | NM_004900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000333467.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000333467.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000333467.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000333467.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000333467.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000333467.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000333467.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000333467.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000333467.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000333467.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000333467.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000333467.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000335760.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000335760.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000335760.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000335760.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000335760.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000335760.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000335760.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000335760.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000335760.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000402182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000402182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000402182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000402182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000402182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000402182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000402182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000402182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000402182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000402182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000402182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000402182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000402182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000407298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000407298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000407298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000407298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000407298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000407298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000407298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000407298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000407298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000407298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | ENST00000407298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | NM_001270411.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | NM_001270411.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | NM_001270411.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | NM_001270411.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | NM_001270411.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | NM_001270411.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | NM_001270411.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | NM_001270411.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | NM_001270411.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | NM_001270411.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | NM_001270411.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | NM_004900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | NM_004900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | NM_004900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | NM_004900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | NM_004900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | NM_004900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | NM_004900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | NM_004900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | NM_004900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | NM_004900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | NM_004900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009270.hg.1 | NM_004900.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200168862.hg.1 | ENST00000333467.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168862.hg.1 | ENST00000335760.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168862.hg.1 | ENST00000402182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168862.hg.1 | ENST00000407298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168862.hg.1 | NM_001270411.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200168862.hg.1 | NM_004900.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200168862.hg.1 | TC2200009270.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200168862.hg.1 | ENST00000333467.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168862.hg.1 | ENST00000335760.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168862.hg.1 | ENST00000402182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168862.hg.1 | ENST00000407298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168862.hg.1 | NM_001270411.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200168862.hg.1 | NM_004900.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200168859.hg.1 | ENST00000333467.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168859.hg.1 | ENST00000335760.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168859.hg.1 | ENST00000402182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168859.hg.1 | ENST00000407298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168859.hg.1 | NM_001270411.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200168859.hg.1 | NM_004900.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200168859.hg.1 | TC2200009270.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200168859.hg.1 | ENST00000333467.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168859.hg.1 | ENST00000335760.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168859.hg.1 | ENST00000402182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168859.hg.1 | ENST00000407298.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168859.hg.1 | NM_001270411.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200168859.hg.1 | NM_004900.4 | Refseq |

APOBEC3B
Genomic Map