Assay Details
| Gene Symbol: CCNT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001240.3 | NP_001231.2 | 2 - 3 | 564 | 137 | |
| NM_001277842.1 | NP_001264771.1 | 2 - 3 | 564 | 137 | ||
| GenBank mRNA | AF045161.1 | - | 2 - 3 | 241 | 137 | |
| AF048730.1 | - | 2 - 3 | 137 | |||
| AK290990.1 | - | 2 - 3 | 464 | 137 | ||
| EF688064.1 | - | 2 - 3 | 241 | 137 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001456.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cyclin-dependent protein serine/threonine kinase activity
transcription from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
protein phosphorylation
cell cycle
viral process
snRNA transcription from RNA polymerase II promoter
positive regulation by host of viral transcription
positive regulation of cyclin-dependent protein serine/threonine kinase activity
positive regulation of transcription from RNA polymerase II promoter
positive regulation of viral transcription
cell division
negative regulation of mRNA polyadenylation
positive regulation of phosphorylation of RNA polymerase II C-terminal domain
DNA binding
chromatin binding
protein serine/threonine kinase activity
protein binding
transcription factor binding
cyclin-dependent protein serine/threonine kinase regulator activity
transcription regulatory region DNA binding
RNA polymerase binding
7SK snRNA binding
regulation of cyclin-dependent protein serine/threonine kinase activity
transcription from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
protein phosphorylation
cell cycle
viral process
snRNA transcription from RNA polymerase II promoter
positive regulation by host of viral transcription
positive regulation of cyclin-dependent protein serine/threonine kinase activity
positive regulation of transcription from RNA polymerase II promoter
positive regulation of viral transcription
cell division
negative regulation of mRNA polyadenylation
positive regulation of phosphorylation of RNA polymerase II C-terminal domain
DNA binding
chromatin binding
protein serine/threonine kinase activity
protein binding
transcription factor binding
cyclin-dependent protein serine/threonine kinase regulator activity
transcription regulatory region DNA binding
RNA polymerase binding
7SK snRNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12019025.hg.1 | ENST00000261900.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12019025.hg.1 | NM_001240.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12019025.hg.1 | TC12001456.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000261900.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000261900.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000261900.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000261900.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000261900.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000261900.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000261900.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000261900.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000261900.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000261900.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000261900.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000261900.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000261900.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000417344.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000417344.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000417344.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000417344.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000417344.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000417344.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000417344.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000417344.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000417344.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000550457.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000550457.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000550457.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000551989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000551989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000551989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000618666.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000618666.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000618666.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000618666.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000618666.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000618666.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000618666.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000618666.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000618666.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000618666.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000618666.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000618666.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | ENST00000618666.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | NM_001240.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | NM_001240.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | NM_001240.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | NM_001240.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | NM_001240.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | NM_001240.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | NM_001240.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | NM_001240.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | NM_001240.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | NM_001240.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | NM_001240.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | NM_001240.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | NM_001240.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | NM_001240.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | NM_001277842.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | NM_001277842.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | NM_001277842.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | NM_001277842.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | NM_001277842.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | NM_001277842.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | NM_001277842.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | NM_001277842.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | NM_001277842.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | NM_001277842.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | NM_001277842.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | NM_001277842.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010591.hg.1 | NM_001277842.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200180159.hg.1 | ENST00000261900.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180159.hg.1 | ENST00000417344.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180159.hg.1 | ENST00000550457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180159.hg.1 | ENST00000618666.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180159.hg.1 | NM_001240.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200180159.hg.1 | NM_001277842.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200180159.hg.1 | ENST00000261900.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180159.hg.1 | ENST00000417344.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180159.hg.1 | ENST00000550457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180159.hg.1 | ENST00000618666.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180159.hg.1 | NM_001240.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200180159.hg.1 | NM_001277842.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200180159.hg.1 | TC1200010591.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000261900.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000261900.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000261900.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000261900.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000261900.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000261900.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000261900.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000261900.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000261900.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000261900.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000261900.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000261900.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000261900.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000417344.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000417344.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000417344.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000417344.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000417344.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000417344.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000417344.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000417344.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000417344.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000550457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000550457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000550457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000551989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000551989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000551989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000618666.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000618666.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000618666.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000618666.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000618666.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000618666.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000618666.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000618666.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000618666.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000618666.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000618666.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000618666.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | ENST00000618666.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | NM_001240.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | NM_001240.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | NM_001240.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | NM_001240.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | NM_001240.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | NM_001240.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | NM_001240.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | NM_001240.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | NM_001240.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | NM_001240.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | NM_001240.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | NM_001240.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | NM_001240.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | NM_001240.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | NM_001277842.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | NM_001277842.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | NM_001277842.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | NM_001277842.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | NM_001277842.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | NM_001277842.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | NM_001277842.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | NM_001277842.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | NM_001277842.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | NM_001277842.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | NM_001277842.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | NM_001277842.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010591.hg.1 | NM_001277842.1 | Refseq |

CCNT1
Genomic Map