Assay Details
| Gene Symbol: LMNB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198557.1 | NP_001185486.1 | 6 - 7 | 890 | 79 | |
| NM_005573.3 | NP_005564.1 | 6 - 7 | 2043 | 79 | ||
| NR_134488.1 | 7 - 8 | 2121 | 79 | |||
| GenBank mRNA | AK303084.1 | - | 6 - 7 | 890 | 79 | |
| AK312603.1 | - | 6 - 7 | 1507 | 79 | ||
| BC012295.1 | - | 6 - 7 | 1441 | 3445475 | 79 | |
| BC103723.1 | - | 6 - 7 | 1538 | 6146212 | 79 | |
| M34458.1 | - | 6 - 7 | 1498 | 79 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000604.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000261366.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000261366.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000261366.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000261366.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000261366.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000261366.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000261366.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000261366.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000261366.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000261366.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000261366.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000261366.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000261366.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000261366.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000261366.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000261366.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000261366.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000261366.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000261366.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000261366.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000261366.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000261366.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000261366.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000395354.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000395354.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000395354.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000395354.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000395354.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000395354.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000395354.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000395354.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000395354.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000395354.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000395354.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000395354.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000395354.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000395354.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000460265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000460265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000460265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000460265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000460265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000460265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000460265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000460265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000460265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000460265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000460265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000460265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000460265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000460265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000460265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000460265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000460265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000460265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000460265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000460265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000460265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000460265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000460265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000460265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000460265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000463908.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000463908.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000463908.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000463908.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000463908.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000472034.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000472034.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000472034.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000472034.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000472034.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000472034.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000472034.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000472034.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000492190.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000492190.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000492190.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000492190.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000492190.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000492190.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000492190.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000494185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000494185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000494185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000494185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000494185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000504788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000504788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000504788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000504788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000504788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000504788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000504788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000504788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000504788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000504788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000504788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000504788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000504788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000504788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000504788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000504788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | ENST00000504788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_001198557.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_001198557.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_001198557.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_001198557.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_001198557.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_001198557.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_001198557.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_001198557.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_001198557.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_001198557.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_001198557.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_001198557.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_001198557.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_001198557.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_001198557.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_001198557.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_001198557.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_001198557.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_005573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_005573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_005573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_005573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_005573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_005573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_005573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_005573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_005573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_005573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_005573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_005573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_005573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_005573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_005573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_005573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_005573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_005573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_005573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_005573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_005573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_005573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_005573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008544.hg.1 | NM_005573.3 | Refseq |

LMNB1
Genomic Map