Assay Details
| Gene Symbol: LRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002332.2 | NP_002323.2 | 87 - 88 | 13813 | 66 | |
| XM_017019303.1 | XP_016874792.1 | 88 - 89 | 13962 | 66 | ||
| GenBank mRNA | AB209498.1 | - | 65 - 66 | 9558 | 66 | |
| AK122840.1 | - | 15 - 16 | 4041 | 66 | ||
| X13916.1 | - | 87 - 88 | 13813 | 66 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000521.hg.1
Set Membership: |
|

LRP1
Genomic Map