Assay Details
| Gene Symbol: TIMMDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016589.3 | NP_057673.2 | 5 - 6 | 809 | 88 | |
| GenBank mRNA | AF139077.1 | - | 5 - 6 | 782 | 88 | |
| AL136622.1 | - | 5 - 6 | 784 | 88 | ||
| AL390090.1 | - | 5 - 6 | 736 | 88 | ||
| AL390094.1 | - | 5 - 6 | 443 | 88 | ||
| BC012341.1 | - | 5 - 6 | 703 | 4576562 | 88 | |
| CR533524.1 | - | 5 - 6 | 596 | 88 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000609.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03011004.hg.1 | ENST00000493694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011004.hg.1 | ENST00000494664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011004.hg.1 | NM_016589.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011004.hg.1 | TC03000609.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000264244.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000264244.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000264244.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000264244.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000264244.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000264244.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000264244.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000264244.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000264244.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000264244.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000264244.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000264244.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000463927.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000463927.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000463927.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000463927.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000463927.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000463927.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000463927.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000466984.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000466984.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000466984.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000466984.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000466984.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000466984.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000466984.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000469324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000469324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000469324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000469324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000486418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000486418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000486418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000486418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000486418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000486418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000492164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000492164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000492164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000492164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000492164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000492164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000492164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000492164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000492164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000493694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000493694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000493694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000493694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000493694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000493694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000493694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000493694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000494664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000494664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000494664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000494664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000494664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000494664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000494664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000494664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000494664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000494664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000494664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000494664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000494664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000494664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000494664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000498399.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000498399.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000498399.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000498399.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000498399.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | ENST00000498399.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | NM_016589.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | NM_016589.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | NM_016589.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | NM_016589.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | NM_016589.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | NM_016589.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | NM_016589.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | NM_016589.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | NM_016589.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | NM_016589.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | NM_016589.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | NM_016589.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | NM_016589.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008477.hg.1 | NM_016589.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300160374.hg.1 | ENST00000264244.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160374.hg.1 | ENST00000463927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160374.hg.1 | ENST00000466984.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160374.hg.1 | ENST00000486418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160374.hg.1 | ENST00000492164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160374.hg.1 | ENST00000493694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160374.hg.1 | ENST00000494664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160374.hg.1 | ENST00000498399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160374.hg.1 | NM_016589.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300160374.hg.1 | ENST00000264244.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160374.hg.1 | ENST00000463927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160374.hg.1 | ENST00000466984.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160374.hg.1 | ENST00000486418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160374.hg.1 | ENST00000498399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160374.hg.1 | ENST00000492164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160374.hg.1 | ENST00000493694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160374.hg.1 | ENST00000494664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160374.hg.1 | NM_016589.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300160374.hg.1 | TC0300008477.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000264244.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000264244.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000264244.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000264244.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000264244.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000264244.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000264244.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000264244.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000264244.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000264244.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000264244.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000264244.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000463927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000463927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000463927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000463927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000463927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000463927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000463927.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000466984.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000466984.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000466984.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000466984.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000466984.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000466984.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000466984.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000469324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000469324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000469324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000469324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000486418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000486418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000486418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000486418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000486418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000486418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000492164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000492164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000492164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000492164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000492164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000492164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000492164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000492164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000492164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000493694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000493694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000493694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000493694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000493694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000493694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000493694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000493694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000494664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000494664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000494664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000494664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000494664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000494664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000494664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000494664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000494664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000494664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000494664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000494664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000494664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000494664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000494664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000498399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000498399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000498399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000498399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000498399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | ENST00000498399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | NM_016589.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | NM_016589.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | NM_016589.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | NM_016589.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | NM_016589.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | NM_016589.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | NM_016589.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | NM_016589.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | NM_016589.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | NM_016589.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | NM_016589.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | NM_016589.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | NM_016589.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008477.hg.1 | NM_016589.3 | Refseq |

TIMMDC1
Genomic Map