Assay Details
| Gene Symbol: ANAPC16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242546.1 | NP_001229475.1 | 3 - 4 | 641 | 100 | |
| NM_001242547.1 | NP_001229476.1 | 2 - 3 | 249 | 100 | ||
| NM_173473.3 | NP_775744.1 | 2 - 3 | 315 | 100 | ||
| GenBank mRNA | BC009530.1 | - | 2 - 3 | 267 | 3890955 | 100 |
| CD511334.1 | - | 2 - 3 | 242 | 100 | ||
| DA739927.1 | - | 2 - 3 | 315 | 100 | ||
| EU552924.1 | - | 2 - 3 | 288 | 100 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000443.hg.1
Set Membership: |
|

ANAPC16
Genomic Map