Assay Details
| Gene Symbol: TYRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000550.2 | NP_000541.1 | 2 - 3 | 561 | 133 | |
| GenBank mRNA | BC052608.1 | - | 1 - 2 | 480 | 6258285 | 133 |
| CR407683.1 | - | 1 - 2 | 383 | 133 | ||
| X51420.1 | - | 2 - 3 | 512 | 133 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000065.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
melanocyte differentiation
melanosome organization
melanin biosynthetic process
acetoacetic acid metabolic process
positive regulation of melanin biosynthetic process
oxidation-reduction process
copper ion binding
protein binding
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, another compound as one donor, and incorporation of one atom of oxygen
protein homodimerization activity
protein heterodimerization activity
melanocyte differentiation
melanosome organization
melanin biosynthetic process
acetoacetic acid metabolic process
positive regulation of melanin biosynthetic process
oxidation-reduction process
copper ion binding
protein binding
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, another compound as one donor, and incorporation of one atom of oxygen
protein homodimerization activity
protein heterodimerization activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000381136.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000381136.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000381136.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000381136.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000381136.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000381136.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000381142.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000381142.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000381142.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000381142.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000381142.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000381142.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000459790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000459790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000459790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000459790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000459790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000459790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000459790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000470909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000470909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000473504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000473504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000473504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000473504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000473504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000473504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000473763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000473763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | ENST00000473763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900006653.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09000774.hg.1 | ENST00000381137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09000774.hg.1 | ENST00000388918.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09000774.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09000774.hg.1 | TC09000065.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09000776.hg.1 | ENST00000381137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09000776.hg.1 | ENST00000388918.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09000776.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09000776.hg.1 | TC09000065.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09000777.hg.1 | ENST00000381137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09000777.hg.1 | ENST00000388918.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09000777.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09000777.hg.1 | TC09000065.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09000773.hg.1 | ENST00000381137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09000773.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09000773.hg.1 | TC09000065.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09000779.hg.1 | ENST00000381137.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09000779.hg.1 | ENST00000388918.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09000779.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09000779.hg.1 | TC09000065.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0900145467.hg.1 | ENST00000459790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900145467.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900145467.hg.1 | ENST00000459790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900145467.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900145467.hg.1 | TC0900006653.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900145470.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900145470.hg.1 | ENST00000459790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900145470.hg.1 | ENST00000473763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900145470.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900145470.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900145470.hg.1 | ENST00000459790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900145470.hg.1 | ENST00000473763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900145470.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900145470.hg.1 | TC0900006653.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900145471.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900145471.hg.1 | ENST00000459790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900145471.hg.1 | ENST00000473763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900145471.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900145471.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900145471.hg.1 | ENST00000459790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900145471.hg.1 | ENST00000473763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900145471.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900145471.hg.1 | TC0900006653.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900145468.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900145468.hg.1 | ENST00000459790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900145468.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900145468.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900145468.hg.1 | ENST00000459790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900145468.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900145468.hg.1 | TC0900006653.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900145474.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900145474.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900145474.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900145474.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900145474.hg.1 | TC0900006653.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000381136.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000381136.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000381136.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000381136.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000381136.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000381136.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000381142.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000381142.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000381142.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000381142.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000381142.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000381142.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000459790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000459790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000459790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000459790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000459790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000459790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000459790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000470909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000470909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000473504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000473504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000473504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000473504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000473504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000473504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000473763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000473763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | ENST00000473763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900006653.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900145472.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900145472.hg.1 | ENST00000459790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900145472.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900145472.hg.1 | ENST00000388918.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900145472.hg.1 | ENST00000459790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900145472.hg.1 | NM_000550.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900145472.hg.1 | TC0900006653.hg.1 | Clariom D TC |

TYRP1
Genomic Map