| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
ENST00000262352.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
ENST00000262352.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
ENST00000262352.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
ENST00000262352.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
ENST00000262352.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
ENST00000262352.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
ENST00000262352.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
ENST00000262352.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
ENST00000262352.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
ENST00000262352.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
ENST00000262352.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
ENST00000262352.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
ENST00000262352.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
ENST00000262352.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
ENST00000262352.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
ENST00000262352.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
ENST00000422398.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
ENST00000422398.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
ENST00000422398.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
ENST00000422398.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
ENST00000422398.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
ENST00000490167.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
ENST00000490167.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
ENST00000490167.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006522.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09000350.hg.1 |
ENST00000262352.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09000350.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09000350.hg.1 |
TC09000029.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09000349.hg.1 |
ENST00000262352.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09000349.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09000349.hg.1 |
TC09000029.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
ENST00000262352.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
ENST00000262352.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
ENST00000262352.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
ENST00000262352.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
ENST00000262352.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
ENST00000262352.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
ENST00000262352.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
ENST00000262352.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
ENST00000262352.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
ENST00000262352.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
ENST00000262352.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
ENST00000262352.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
ENST00000262352.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
ENST00000262352.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
ENST00000262352.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
ENST00000262352.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
ENST00000422398.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
ENST00000422398.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
ENST00000422398.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
ENST00000422398.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
ENST00000422398.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
ENST00000490167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
ENST00000490167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
ENST00000490167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006522.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900144701.hg.1 |
ENST00000262352.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900144701.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900144701.hg.1 |
ENST00000262352.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900144701.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900144701.hg.1 |
TC0900006522.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900144702.hg.1 |
ENST00000262352.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900144702.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900144702.hg.1 |
ENST00000262352.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900144702.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900144702.hg.1 |
TC0900006522.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900144703.hg.1 |
ENST00000262352.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900144703.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900144703.hg.1 |
ENST00000262352.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900144703.hg.1 |
NM_004170.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900144703.hg.1 |
TC0900006522.hg.1 |
Clariom D TC |