Assay Details
| Gene Symbol: GARNL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104591.1 | 24 - 25 | 2788 | 108 | ||
| GenBank mRNA | AL136573.1 | - | 24 - 25 | 2342 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GARNL3| Gene Symbol: GARNL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104591.1 | 24 - 25 | 2788 | 108 | ||
| GenBank mRNA | AL136573.1 | - | 24 - 25 | 2342 | 108 | |
Set Membership: |
|