| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04016006.hg.1 |
ENST00000381571.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04016006.hg.1 |
NM_017845.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04016006.hg.1 |
TC04001167.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04016007.hg.1 |
ENST00000381571.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04016007.hg.1 |
NM_017845.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04016007.hg.1 |
TC04001167.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010606.hg.1 |
ENST00000381571.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010606.hg.1 |
ENST00000381571.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010606.hg.1 |
ENST00000381571.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010606.hg.1 |
ENST00000381571.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010606.hg.1 |
ENST00000381571.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010606.hg.1 |
ENST00000381571.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010606.hg.1 |
ENST00000381571.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010606.hg.1 |
ENST00000509220.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010606.hg.1 |
ENST00000509220.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010606.hg.1 |
ENST00000509220.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010606.hg.1 |
NM_017845.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010606.hg.1 |
NM_017845.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010606.hg.1 |
NM_017845.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010606.hg.1 |
NM_017845.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010606.hg.1 |
NM_017845.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010606.hg.1 |
NM_017845.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400172295.hg.1 |
ENST00000381571.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400172295.hg.1 |
ENST00000381571.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400172295.hg.1 |
TC0400010606.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0400010606.hg.1 |
ENST00000381571.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010606.hg.1 |
ENST00000381571.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010606.hg.1 |
ENST00000381571.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010606.hg.1 |
ENST00000381571.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010606.hg.1 |
ENST00000381571.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010606.hg.1 |
ENST00000381571.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010606.hg.1 |
ENST00000381571.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010606.hg.1 |
ENST00000509220.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010606.hg.1 |
ENST00000509220.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010606.hg.1 |
ENST00000509220.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010606.hg.1 |
NM_017845.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010606.hg.1 |
NM_017845.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010606.hg.1 |
NM_017845.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010606.hg.1 |
NM_017845.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010606.hg.1 |
NM_017845.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010606.hg.1 |
NM_017845.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400172291.hg.1 |
ENST00000381571.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400172291.hg.1 |
ENST00000509220.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400172291.hg.1 |
NM_017845.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400172291.hg.1 |
ENST00000381571.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400172291.hg.1 |
ENST00000509220.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400172291.hg.1 |
NM_017845.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400172291.hg.1 |
TC0400010606.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400172290.hg.1 |
ENST00000381571.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400172290.hg.1 |
ENST00000509220.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400172290.hg.1 |
NM_017845.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400172290.hg.1 |
ENST00000381571.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400172290.hg.1 |
ENST00000509220.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400172290.hg.1 |
NM_017845.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400172290.hg.1 |
TC0400010606.hg.1 |
Clariom D TC |