Assay Details
| Gene Symbol: PTH2R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309516.1 | NP_001296445.1 | 4 - 5 | 732 | 101 | |
| NM_005048.3 | NP_005039.1 | 4 - 5 | 628 | 101 | ||
| GenBank mRNA | AK294181.1 | - | 4 - 5 | 732 | 101 | |
| AY497546.1 | - | 4 - 5 | 414 | 101 | ||
| BC036811.1 | - | 4 - 5 | 627 | 5734330 | 101 | |
| BM808818.1 | - | 4 - 5 | 640 | 101 | ||
| U25128.1 | - | 4 - 5 | 556 | 101 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001236.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000487472.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000487472.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000487472.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000487472.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000487472.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000487472.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02018955.hg.1 | ENST00000272847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02018955.hg.1 | ENST00000413482.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02018955.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02018955.hg.1 | TC02001236.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000487472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000487472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000487472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000487472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000487472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000487472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175320.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200175320.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200175320.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175320.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175320.hg.1 | ENST00000272847.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200175320.hg.1 | ENST00000617735.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200175320.hg.1 | NM_001309516.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175320.hg.1 | NM_005048.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175320.hg.1 | TC0200010627.hg.1 | Clariom D TC |

PTH2R
Genomic Map