Assay Details
| Gene Symbol: DUSP11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003584.2 | NP_003575.2 | 1 - 2 | 289 | 63 | |
| GenBank mRNA | AF023917.1 | - | 1 - 2 | 230 | 63 | |
| AK315271.1 | - | 1 - 2 | 241 | 63 | ||
| BC000346.2 | - | 1 - 2 | 207 | 2960939 | 63 | |
| BP349009.1 | - | 1 - 2 | 241 | 63 | ||
| BP359552.1 | - | 1 - 2 | 289 | 63 | ||
| CR627368.1 | - | 1 - 2 | 253 | 63 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001988.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
RNA processing
protein dephosphorylation
RNA metabolic process
peptidyl-tyrosine dephosphorylation
polynucleotide 5' dephosphorylation
RNA binding
polynucleotide 5'-phosphatase activity
protein tyrosine phosphatase activity
protein tyrosine/serine/threonine phosphatase activity
phosphatase activity
poly(A) RNA binding
nucleotide phosphatase activity, acting on free nucleotides
RNA processing
protein dephosphorylation
RNA metabolic process
peptidyl-tyrosine dephosphorylation
polynucleotide 5' dephosphorylation
RNA binding
polynucleotide 5'-phosphatase activity
protein tyrosine phosphatase activity
protein tyrosine/serine/threonine phosphatase activity
phosphatase activity
poly(A) RNA binding
nucleotide phosphatase activity, acting on free nucleotides
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02031695.hg.1 | ENST00000272444.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031695.hg.1 | ENST00000377706.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031695.hg.1 | ENST00000443070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031695.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031695.hg.1 | TC02001988.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000443070.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000443070.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000443070.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000443070.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000443070.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000443070.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000443070.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000443070.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000452812.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000452812.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000452812.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000452812.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000452812.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000477458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000477458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000477458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000477458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000477458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000477458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000477458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000480948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000480948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000480948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000480948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000480948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000480948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000480948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000480948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | ENST00000480948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013079.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031692.hg.1 | ENST00000272444.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031692.hg.1 | ENST00000377706.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031692.hg.1 | ENST00000443070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031692.hg.1 | ENST00000480948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031692.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031692.hg.1 | TC02001988.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031696.hg.1 | ENST00000272444.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031696.hg.1 | ENST00000377706.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031696.hg.1 | ENST00000443070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031696.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031696.hg.1 | TC02001988.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031698.hg.1 | ENST00000272444.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031698.hg.1 | ENST00000443070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031698.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031698.hg.1 | TC02001988.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031699.hg.1 | ENST00000272444.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031699.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031699.hg.1 | TC02001988.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0200195497.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195497.hg.1 | ENST00000443070.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195497.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195497.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195497.hg.1 | ENST00000443070.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195497.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195497.hg.1 | TC0200013079.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000443070.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000443070.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000443070.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000443070.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000443070.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000443070.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000443070.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000443070.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000452812.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000452812.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000452812.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000452812.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000452812.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000477458.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000477458.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000477458.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000477458.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000477458.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000477458.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000477458.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000480948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000480948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000480948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000480948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000480948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000480948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000480948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000480948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | ENST00000480948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013079.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195498.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195498.hg.1 | ENST00000443070.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195498.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195498.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195498.hg.1 | ENST00000443070.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195498.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195498.hg.1 | TC0200013079.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200195496.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195496.hg.1 | ENST00000443070.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195496.hg.1 | ENST00000452812.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195496.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195496.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195496.hg.1 | ENST00000443070.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195496.hg.1 | ENST00000452812.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195496.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195496.hg.1 | TC0200013079.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200195500.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195500.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195500.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195500.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195500.hg.1 | TC0200013079.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200195493.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195493.hg.1 | ENST00000443070.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195493.hg.1 | ENST00000452812.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195493.hg.1 | ENST00000477458.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195493.hg.1 | ENST00000480948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195493.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195493.hg.1 | ENST00000272444.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195493.hg.1 | ENST00000443070.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195493.hg.1 | ENST00000452812.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195493.hg.1 | ENST00000477458.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195493.hg.1 | ENST00000480948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195493.hg.1 | NM_003584.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195493.hg.1 | TC0200013079.hg.1 | Clariom D TC |

DUSP11
Genomic Map