Assay Details
| Gene Symbol: CBR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286789.1 | NP_001273718.1 | 2 - 3 | 587 | 84 | |
| GenBank mRNA | AK294142.1 | - | 2 - 3 | 488 | 84 | |
| AK311219.1 | - | 2 - 3 | 487 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
drug metabolic process
cyclooxygenase pathway
epithelial cell differentiation
vitamin K metabolic process
oxidation-reduction process
carbonyl reductase (NADPH) activity
oxidoreductase activity, acting on NAD(P)H, quinone or similar compound as acceptor
15-hydroxyprostaglandin dehydrogenase (NADP+) activity
prostaglandin-E2 9-reductase activity
drug metabolic process
cyclooxygenase pathway
epithelial cell differentiation
vitamin K metabolic process
oxidation-reduction process
carbonyl reductase (NADPH) activity
oxidoreductase activity, acting on NAD(P)H, quinone or similar compound as acceptor
15-hydroxyprostaglandin dehydrogenase (NADP+) activity
prostaglandin-E2 9-reductase activity
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000399191.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000399191.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000399191.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000399191.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000399191.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000439427.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000439427.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000439427.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000439427.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000439427.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000466328.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000466328.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000466328.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000466328.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | NM_001757.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | NM_001757.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | NM_001757.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | NM_001757.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | NM_001757.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | NM_001757.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | NM_001757.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007032.hg.1 | NM_001757.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100147455.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147455.hg.1 | ENST00000399191.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147455.hg.1 | ENST00000439427.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147455.hg.1 | ENST00000466328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147455.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147455.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100147455.hg.1 | NM_001757.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100147455.hg.1 | TC2100007032.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100147455.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147455.hg.1 | ENST00000399191.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147455.hg.1 | ENST00000439427.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147455.hg.1 | ENST00000466328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147455.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147455.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100147455.hg.1 | NM_001757.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000399191.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000399191.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000399191.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000399191.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000399191.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000439427.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000439427.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000439427.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000439427.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000439427.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000466328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000466328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000466328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000466328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | NM_001757.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | NM_001757.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | NM_001757.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | NM_001757.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | NM_001757.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | NM_001757.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | NM_001757.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007032.hg.1 | NM_001757.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100147459.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147459.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100147459.hg.1 | TC2100007032.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100147459.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147459.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100147460.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147460.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100147460.hg.1 | TC2100007032.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100147460.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147460.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100147453.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147453.hg.1 | ENST00000399191.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147453.hg.1 | ENST00000439427.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147453.hg.1 | ENST00000466328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147453.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147453.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100147453.hg.1 | NM_001757.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100147453.hg.1 | TC2100007032.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100147453.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147453.hg.1 | ENST00000399191.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147453.hg.1 | ENST00000439427.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147453.hg.1 | ENST00000466328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147453.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147453.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100147453.hg.1 | NM_001757.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100147449.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147449.hg.1 | ENST00000466328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147449.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147449.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100147449.hg.1 | NM_001757.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100147449.hg.1 | TC2100007032.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100147449.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147449.hg.1 | ENST00000466328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147449.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147449.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100147449.hg.1 | NM_001757.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100147461.hg.1 | ENST00000399191.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147461.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147461.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100147461.hg.1 | TC2100007032.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100147461.hg.1 | ENST00000399191.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147461.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147461.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100147454.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147454.hg.1 | ENST00000399191.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147454.hg.1 | ENST00000439427.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147454.hg.1 | ENST00000466328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147454.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147454.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100147454.hg.1 | NM_001757.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100147454.hg.1 | TC2100007032.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100147454.hg.1 | ENST00000290349.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147454.hg.1 | ENST00000399191.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147454.hg.1 | ENST00000439427.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147454.hg.1 | ENST00000466328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147454.hg.1 | ENST00000530908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147454.hg.1 | NM_001286789.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100147454.hg.1 | NM_001757.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 214556 | - | - |

CBR1
Genomic Map