Assay Details
| Gene Symbol: MTFP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016498.4 | NP_057582.2 | 2 - 3 | 452 | 112 | |
| GenBank mRNA | AF151076.1 | - | 2 - 3 | 414 | 112 | |
| AK026104.1 | - | 2 - 3 | 320 | 112 | ||
| BC001608.1 | - | 2 - 3 | 278 | 3542567 | 112 | |
| BC009300.2 | - | 2 - 3 | 310 | 4126237 | 112 | |
| BC014446.1 | - | 2 - 3 | 278 | 4855192 | 112 | |
| BC030989.2 | - | 2 - 3 | 339 | 4077757 | 112 | |
| BC038831.2 | - | 2 - 3 | 343 | 3908299 | 112 | |
| BC046132.1 | - | 2 - 3 | 242 | 4446034 | 112 | |
| CR456345.1 | - | 2 - 3 | 247 | 112 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22001462.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000266263.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000266263.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000266263.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000266263.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000266263.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000266263.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000266263.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000266263.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000266263.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000266263.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000266263.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000266263.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000355143.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000355143.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000355143.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000355143.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000355143.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000355143.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000355143.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000355143.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000407550.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000407550.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000407550.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000407550.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000407550.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000407550.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000407550.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000407550.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000412752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000412752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000412752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000412752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000412752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000412752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000412752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000412752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000614920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000614920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000614920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000614920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000614920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000629597.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000629597.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000629597.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000629597.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | ENST00000629597.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | NM_001003704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | NM_001003704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | NM_001003704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | NM_001003704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | NM_001003704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | NM_001003704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | NM_001003704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | NM_001003704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | NM_001003704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | NM_016498.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | NM_016498.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | NM_016498.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | NM_016498.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | NM_016498.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | NM_016498.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | NM_016498.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | NM_016498.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | NM_016498.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | NM_016498.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009253.hg.1 | NM_016498.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22003989.hg.1 | ENST00000266263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22003989.hg.1 | ENST00000407550.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22003989.hg.1 | NM_016498.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22003989.hg.1 | TC22001462.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22003988.hg.1 | ENST00000266263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22003988.hg.1 | NM_016498.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22003988.hg.1 | TC22001462.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2200168487.hg.1 | ENST00000266263.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168487.hg.1 | ENST00000412752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168487.hg.1 | NM_016498.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200168487.hg.1 | TC2200009253.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200168487.hg.1 | ENST00000266263.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168487.hg.1 | ENST00000412752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168487.hg.1 | NM_016498.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200168488.hg.1 | ENST00000266263.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168488.hg.1 | ENST00000407550.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168488.hg.1 | ENST00000412752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168488.hg.1 | NM_016498.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200168488.hg.1 | TC2200009253.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200168488.hg.1 | ENST00000266263.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168488.hg.1 | ENST00000407550.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168488.hg.1 | ENST00000412752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200168488.hg.1 | NM_016498.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000266263.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000266263.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000266263.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000266263.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000266263.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000266263.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000266263.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000266263.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000266263.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000266263.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000266263.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000266263.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000355143.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000355143.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000355143.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000355143.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000355143.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000355143.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000355143.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000355143.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000407550.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000407550.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000407550.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000407550.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000407550.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000407550.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000407550.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000407550.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000412752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000412752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000412752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000412752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000412752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000412752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000412752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000412752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000614920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000614920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000614920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000614920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000614920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000629597.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000629597.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000629597.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000629597.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | ENST00000629597.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | NM_001003704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | NM_001003704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | NM_001003704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | NM_001003704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | NM_001003704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | NM_001003704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | NM_001003704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | NM_001003704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | NM_001003704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | NM_016498.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | NM_016498.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | NM_016498.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | NM_016498.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | NM_016498.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | NM_016498.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | NM_016498.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | NM_016498.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | NM_016498.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | NM_016498.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009253.hg.1 | NM_016498.4 | Refseq |

MTFP1
Genomic Map