| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17010866.hg.1 |
ENST00000301585.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17010866.hg.1 |
NM_001545.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17010866.hg.1 |
TC17000839.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
ENST00000301585.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
ENST00000301585.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
ENST00000301585.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
ENST00000301585.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
ENST00000301585.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
ENST00000301585.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
ENST00000301585.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
ENST00000301585.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
ENST00000301585.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
ENST00000580800.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
ENST00000580800.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
ENST00000580800.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
ENST00000580800.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
ENST00000580800.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
ENST00000580800.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
ENST00000583772.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
ENST00000583772.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
ENST00000583772.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
ENST00000583772.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
ENST00000583772.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
ENST00000583772.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
ENST00000584208.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
ENST00000584208.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
ENST00000584208.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
ENST00000584208.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
ENST00000584208.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
ENST00000584208.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
ENST00000584208.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
NM_001303265.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
NM_001303265.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
NM_001303265.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
NM_001303265.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
NM_001303265.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
NM_001303265.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
NM_001303265.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
NM_001303265.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
NM_001303265.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
NM_001545.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
NM_001545.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
NM_001545.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
NM_001545.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
NM_001545.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
NM_001545.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
NM_001545.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
NM_001545.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008879.hg.1 |
NM_001545.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700166349.hg.1 |
ENST00000301585.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166349.hg.1 |
ENST00000580800.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166349.hg.1 |
ENST00000584208.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166349.hg.1 |
NM_001303265.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700166349.hg.1 |
NM_001545.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700166349.hg.1 |
TC1700008879.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700166349.hg.1 |
ENST00000301585.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166349.hg.1 |
ENST00000580800.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166349.hg.1 |
ENST00000584208.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166349.hg.1 |
NM_001303265.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700166349.hg.1 |
NM_001545.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
ENST00000301585.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
ENST00000301585.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
ENST00000301585.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
ENST00000301585.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
ENST00000301585.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
ENST00000301585.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
ENST00000301585.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
ENST00000301585.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
ENST00000301585.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
ENST00000580800.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
ENST00000580800.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
ENST00000580800.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
ENST00000580800.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
ENST00000580800.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
ENST00000580800.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
ENST00000583772.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
ENST00000583772.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
ENST00000583772.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
ENST00000583772.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
ENST00000583772.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
ENST00000583772.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
ENST00000584208.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
ENST00000584208.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
ENST00000584208.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
ENST00000584208.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
ENST00000584208.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
ENST00000584208.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
ENST00000584208.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
NM_001303265.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
NM_001303265.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
NM_001303265.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
NM_001303265.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
NM_001303265.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
NM_001303265.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
NM_001303265.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
NM_001303265.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
NM_001303265.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
NM_001545.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
NM_001545.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
NM_001545.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
NM_001545.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
NM_001545.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
NM_001545.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
NM_001545.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
NM_001545.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008879.hg.1 |
NM_001545.2 |
Refseq |