Assay Details
| Gene Symbol: FSTL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015082.1 | NP_055897.1 | 1 - 2 | 205 | 71 | |
| XM_011543284.1 | XP_011541586.1 | 1 - 2 | 183 | 71 | ||
| GenBank mRNA | AK055684.1 | - | 1 - 2 | 183 | 71 | |
| BC024300.1 | - | 1 - 2 | 237 | 4662520 | 71 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001780.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC024300.1,NM_015082.1,XM_011543284.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000507112.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000507112.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000507112.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000507112.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000507112.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000507112.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000510685.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000510685.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000510685.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000510685.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000510685.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000511375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000511375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000511375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000511375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000514998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000514998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000514998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000514998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000514998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05024211.hg.1 | ENST00000265342.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05024211.hg.1 | ENST00000360575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05024211.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05024211.hg.1 | TC05001780.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05024210.hg.1 | ENST00000265342.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05024210.hg.1 | ENST00000360575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05024210.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05024210.hg.1 | TC05001780.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0500183295.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183295.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500183295.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183295.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500183295.hg.1 | TC0500012044.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500183296.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183296.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183296.hg.1 | TC0500012044.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000507112.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000507112.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000507112.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000507112.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000507112.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000507112.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000509525.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000510685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000510685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000510685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000510685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000510685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000511375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000511375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000511375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000511375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000514998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000514998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000514998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000514998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000514998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012044.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500183292.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183292.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183292.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500183292.hg.1 | ENST00000265342.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183292.hg.1 | ENST00000621681.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500183292.hg.1 | NM_015082.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500183292.hg.1 | TC0500012044.hg.1 | Clariom D TC |

FSTL4
Genomic Map