Assay Details
| Gene Symbol: EVC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306090.1 | NP_001293019.1 | 20 - 21 | 3075 | 83 | |
| GenBank mRNA | AF239742.1 | - | 20 - 21 | 2891 | 83 | |
More Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000506240.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000506240.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000506240.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000506240.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000509451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000509451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000509451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000509451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000509451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000509451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000509451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000509451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000509451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000509451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000509451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000509451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000509451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000514919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000514919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000514919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000515113.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000515113.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000515113.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | ENST00000515113.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_001306092.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_001306092.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_001306092.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_001306092.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_001306092.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_001306092.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_001306092.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_001306092.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_001306092.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_001306092.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_001306092.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_001306092.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_001306092.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000264956.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000506240.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000506240.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000506240.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000506240.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000509451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000509451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000509451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000509451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000509451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000509451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000509451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000509451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000509451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000509451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000509451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000509451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000509451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000514919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000514919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000514919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000515113.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000515113.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000515113.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | ENST00000515113.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_001306090.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_001306092.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_001306092.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_001306092.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_001306092.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_001306092.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_001306092.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_001306092.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_001306092.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_001306092.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_001306092.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_001306092.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_001306092.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_001306092.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_153717.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012760.hg.1 | NM_153717.2 | Refseq |

EVC
Genomic Map