Assay Details
| Gene Symbol: NUBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001323595.1 | NP_001310524.1 | 4 - 5 | 329 | 139 | |
| NM_001323596.1 | NP_001310525.1 | 4 - 5 | 354 | 139 | ||
| NM_001323597.1 | NP_001310526.1 | 3 - 4 | 320 | 139 | ||
| NM_002484.3 | NP_002475.2 | 4 - 5 | 403 | 139 | ||
| GenBank mRNA | AK223204.1 | - | 4 - 5 | 345 | 139 | |
| AK310089.1 | - | 4 - 5 | 348 | 139 | ||
| BC109322.1 | - | 4 - 5 | 329 | 40036841 | 139 | |
| BC109323.1 | - | 4 - 5 | 329 | 40036842 | 139 | |
| BP216107.1 | - | 4 - 5 | 348 | 139 | ||
| U01833.1 | - | 4 - 5 | 329 | 139 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000157.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
cellular iron ion homeostasis
negative regulation of centrosome duplication
cell growth
iron-sulfur cluster assembly
cell projection organization
centrosome localization
protein localization to cell cortex
nucleotide binding
protein binding
ATP binding
metal ion binding
iron-sulfur cluster binding
4 iron, 4 sulfur cluster binding
cellular iron ion homeostasis
negative regulation of centrosome duplication
cell growth
iron-sulfur cluster assembly
cell projection organization
centrosome localization
protein localization to cell cortex
nucleotide binding
protein binding
ATP binding
metal ion binding
iron-sulfur cluster binding
4 iron, 4 sulfur cluster binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000283027.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000283027.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000283027.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000283027.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000283027.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000283027.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000283027.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000283027.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000283027.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000283027.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000283027.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000283027.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000283027.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000433392.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000433392.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000433392.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000433392.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000433392.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000433392.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000433392.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000433392.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000433392.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000433392.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000433392.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000433392.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000571175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000571175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000571175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000571175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000571175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000571175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000571175.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000571790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000571790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000571790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000571790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000571790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000571790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000571790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000571790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000571790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000573663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000573663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000573663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000573663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000573663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000574137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000574137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000574137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000574137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000574137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000574137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000574137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000574334.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000574334.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000574334.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000574334.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000574334.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000574334.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000574691.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000574691.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000574691.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000574691.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000574691.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | ENST00000574691.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | NM_001278506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | NM_001278506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | NM_001278506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | NM_001278506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | NM_001278506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | NM_001278506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | NM_001278506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | NM_001278506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | NM_001278506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | NM_001278506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | NM_001278506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | NM_001278506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | NM_001278506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | NM_002484.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | NM_002484.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | NM_002484.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | NM_002484.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | NM_002484.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | NM_002484.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | NM_002484.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | NM_002484.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | NM_002484.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | NM_002484.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | NM_002484.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | NM_002484.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | NM_002484.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006886.hg.1 | NM_002484.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16002385.hg.1 | ENST00000283027.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16002385.hg.1 | ENST00000433392.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16002385.hg.1 | NM_002484.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16002385.hg.1 | TC16000157.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000283027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000283027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000283027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000283027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000283027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000283027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000283027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000283027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000283027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000283027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000283027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000283027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000283027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000433392.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000433392.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000433392.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000433392.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000433392.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000433392.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000433392.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000433392.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000433392.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000433392.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000433392.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000433392.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000571175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000571175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000571175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000571175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000571175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000571175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000571175.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000571790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000571790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000571790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000571790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000571790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000571790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000571790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000571790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000571790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000573663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000573663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000573663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000573663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000573663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000574137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000574137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000574137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000574137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000574137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000574137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000574137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000574334.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000574334.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000574334.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000574334.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000574334.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000574334.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000574691.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000574691.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000574691.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000574691.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000574691.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | ENST00000574691.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | NM_001278506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | NM_001278506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | NM_001278506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | NM_001278506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | NM_001278506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | NM_001278506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | NM_001278506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | NM_001278506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | NM_001278506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | NM_001278506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | NM_001278506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | NM_001278506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | NM_001278506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | NM_002484.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | NM_002484.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | NM_002484.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | NM_002484.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | NM_002484.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | NM_002484.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | NM_002484.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | NM_002484.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | NM_002484.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | NM_002484.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | NM_002484.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | NM_002484.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | NM_002484.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006886.hg.1 | NM_002484.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600148936.hg.1 | ENST00000283027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148936.hg.1 | ENST00000571790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148936.hg.1 | ENST00000574334.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148936.hg.1 | NM_002484.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600148936.hg.1 | TC1600006886.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600148936.hg.1 | ENST00000283027.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148936.hg.1 | ENST00000571790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148936.hg.1 | ENST00000574334.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600148936.hg.1 | NM_002484.3 | Refseq |

NUBP1
Genomic Map