Assay Details
| Gene Symbol: TCF7L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031283.2 | NP_112573.1 | 1 - 2 | 473 | 89 | |
| XM_006712109.2 | XP_006712172.1 | 1 - 2 | 263 | 89 | ||
| GenBank mRNA | AB031046.1 | - | 1 - 2 | 322 | 89 | |
| BC058894.1 | - | 1 - 2 | 482 | 6141641 | 89 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000502.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
chromatin organization
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
regulation of Wnt signaling pathway
canonical Wnt signaling pathway
beta-catenin-TCF complex assembly
DNA binding
transcription factor activity, sequence-specific DNA binding
beta-catenin binding
sequence-specific DNA binding
transcription regulatory region DNA binding
chromatin organization
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
regulation of Wnt signaling pathway
canonical Wnt signaling pathway
beta-catenin-TCF complex assembly
DNA binding
transcription factor activity, sequence-specific DNA binding
beta-catenin binding
sequence-specific DNA binding
transcription regulatory region DNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000442813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000442813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000442813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000442813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000442813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000490744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000490744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000490744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000490744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000494519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000494519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007693.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007693.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007693.hg.1 | TC02000502.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007695.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007695.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007695.hg.1 | TC02000502.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007696.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007696.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007696.hg.1 | TC02000502.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0200157005.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200157005.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200157005.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200157005.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200157005.hg.1 | TC0200008236.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200157003.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200157003.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200157003.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200157003.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200157003.hg.1 | TC0200008236.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000442813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000442813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000442813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000442813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000442813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000490744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000490744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000490744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000490744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000494519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000494519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200157004.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200157004.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200157004.hg.1 | ENST00000282111.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200157004.hg.1 | NM_031283.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200157004.hg.1 | TC0200008236.hg.1 | Clariom D TC |

TCF7L1
Genomic Map