Assay Details
| Gene Symbol: ANKH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054027.4 | NP_473368.1 | 4 - 5 | 850 | 68 | |
| XM_011514067.1 | XP_011512369.1 | 4 - 5 | 813 | 68 | ||
| XM_017009644.1 | XP_016865133.1 | 4 - 5 | 450 | 68 | ||
| GenBank mRNA | AB046801.1 | - | 4 - 5 | 680 | 68 | |
| AF274753.1 | - | 4 - 5 | 587 | 68 | ||
| AK222631.1 | - | 548 | 68 | |||
| AK315012.1 | - | 4 - 5 | 548 | 68 | ||
| AY358503.1 | - | 4 - 5 | 601 | 68 | ||
| BC009835.2 | - | 4 - 5 | 775 | 3927236 | 68 | |
| BC014526.2 | - | 4 - 5 | 575 | 3837372 | 68 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001172.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
skeletal system development
locomotory behavior
regulation of bone mineralization
inorganic diphosphate transport
phosphate ion transmembrane transport
transmembrane transport
inorganic phosphate transmembrane transporter activity
phosphate ion transmembrane transporter activity
inorganic diphosphate transmembrane transporter activity
skeletal system development
locomotory behavior
regulation of bone mineralization
inorganic diphosphate transport
phosphate ion transmembrane transport
transmembrane transport
inorganic phosphate transmembrane transporter activity
phosphate ion transmembrane transporter activity
inorganic diphosphate transmembrane transporter activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05016818.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016818.hg.1 | ENST00000503939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016818.hg.1 | ENST00000535119.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016818.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016818.hg.1 | TC05001172.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016819.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016819.hg.1 | ENST00000503939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016819.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016819.hg.1 | TC05001172.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000502585.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000502585.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000502585.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000502585.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000502585.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000502585.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000503389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000503389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000503939.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000503939.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000503939.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000503939.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000503939.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000503939.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000503939.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000505140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000505140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000505140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000505140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000505140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000505140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000513115.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000513115.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000513115.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000513115.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000513115.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000515517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000515517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000515517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | ENST00000515517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016821.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016821.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016821.hg.1 | TC05001172.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016820.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016820.hg.1 | ENST00000503939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016820.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016820.hg.1 | TC05001172.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0500171225.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500171225.hg.1 | ENST00000503939.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500171225.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500171225.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500171225.hg.1 | ENST00000503939.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500171225.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500171225.hg.1 | TC0500010169.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500171227.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500171227.hg.1 | ENST00000503939.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500171227.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500171227.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500171227.hg.1 | ENST00000503939.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500171227.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500171227.hg.1 | TC0500010169.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500171228.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500171228.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500171228.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500171228.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500171228.hg.1 | TC0500010169.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000284268.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000502585.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000502585.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000502585.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000502585.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000502585.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000502585.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000503389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000503389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000503939.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000503939.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000503939.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000503939.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000503939.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000503939.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000503939.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000505140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000505140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000505140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000505140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000505140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000505140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000513115.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000513115.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000513115.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000513115.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000513115.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000515517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000515517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000515517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | ENST00000515517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | NM_054027.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010169.hg.1 | NM_054027.5 | Refseq |

ANKH
Genomic Map