Assay Details
| Gene Symbol: TMEM87A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110503.2 | NP_001103973.1 | 5 - 6 | 650 | 88 | |
| GenBank mRNA | BC005335.1 | - | 5 - 6 | 502 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

TMEM87A
Genomic Map