Assay Details
| Gene Symbol: GABPB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320915.1 | NP_001307844.1 | 7 - 8 | 1199 | 75 | |
| XM_011521426.2 | XP_011519728.1 | 6 - 7 | 1118 | 75 | ||
| XM_017022054.1 | XP_016877543.1 | 7 - 8 | 858 | 75 | ||
| GenBank mRNA | D13316.1 | - | 7 - 8 | 835 | 75 | |
| JQ313901.1 | - | 5 - 6 | 694 | 75 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

GABPB1
Genomic Map