Assay Details
| Gene Symbol: TM4SF19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204897.1 | NP_001191826.1 | 2 - 3 | 362 | 89 | |
| NM_138461.3 | NP_612470.2 | 2 - 3 | 362 | 89 | ||
| GenBank mRNA | AF192910.1 | - | 2 - 3 | 318 | 89 | |
| AK310209.1 | - | 2 - 3 | 332 | 89 | ||
| BC146985.1 | - | 2 - 3 | 263 | 9056913 | 89 | |
| BC146993.1 | - | 2 - 3 | 263 | 9056921 | 89 | |
| BC147021.1 | - | 2 - 3 | 263 | 9056949 | 89 | |
| BQ678650.1 | - | 2 - 3 | 328 | 89 | ||
| DB015357.1 | - | 2 - 3 | 332 | 89 | ||
| Gene Symbol: TM4SF19-TCTEX1D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037950.1 | 2 - 3 | 362 | 89 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03003402.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03038224.hg.1 | ENST00000273695.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038224.hg.1 | ENST00000446879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038224.hg.1 | NM_001204897.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038224.hg.1 | NM_138461.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038224.hg.1 | TC03003402.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | ENST00000273695.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | ENST00000273695.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | ENST00000273695.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | ENST00000273695.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | ENST00000273695.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | ENST00000273695.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | ENST00000273695.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | ENST00000273695.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | ENST00000273695.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | ENST00000273695.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | ENST00000440822.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | ENST00000440822.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | ENST00000440822.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | ENST00000440822.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | ENST00000440822.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | ENST00000446879.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | ENST00000446879.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | ENST00000446879.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | ENST00000446879.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | ENST00000446879.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | ENST00000446879.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | ENST00000446879.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | ENST00000446879.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | ENST00000446879.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | ENST00000446879.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | ENST00000454715.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | ENST00000454715.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | ENST00000454715.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | ENST00000454715.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | ENST00000454715.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | ENST00000454715.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | ENST00000454715.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | ENST00000454715.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | ENST00000454715.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | ENST00000454715.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | ENST00000454715.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | NM_001204897.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | NM_001204897.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | NM_001204897.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | NM_001204897.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | NM_001204897.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | NM_001204897.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | NM_001204897.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | NM_001204897.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | NM_001204897.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | NM_001204897.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | NM_001204897.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | NM_001204898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | NM_001204898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | NM_001204898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | NM_001204898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | NM_001204898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | NM_001204898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | NM_001204898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | NM_001204898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | NM_001204898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | NM_001204898.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | NM_138461.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | NM_138461.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | NM_138461.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | NM_138461.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | NM_138461.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | NM_138461.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | NM_138461.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | NM_138461.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | NM_138461.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | NM_138461.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014095.hg.1 | NM_138461.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014092.hg.1 | ENST00000442633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014092.hg.1 | ENST00000442633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014092.hg.1 | ENST00000442633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014092.hg.1 | ENST00000442633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014092.hg.1 | ENST00000442633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014092.hg.1 | ENST00000442633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014092.hg.1 | NR_037950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014092.hg.1 | NR_037950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014092.hg.1 | NR_037950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014092.hg.1 | NR_037950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014092.hg.1 | NR_037950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014092.hg.1 | NR_037950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014092.hg.1 | NR_037950.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300210027.hg.1 | ENST00000273695.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300210027.hg.1 | ENST00000446879.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300210027.hg.1 | NM_001204897.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300210027.hg.1 | NM_138461.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300210027.hg.1 | ENST00000273695.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300210027.hg.1 | ENST00000446879.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300210027.hg.1 | NM_001204897.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300210027.hg.1 | NM_138461.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300210027.hg.1 | TC0300014095.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300209939.hg.1 | ENST00000442633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300209939.hg.1 | NR_037950.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300209939.hg.1 | ENST00000442633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300209939.hg.1 | NR_037950.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300209939.hg.1 | TC0300014092.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300014092.hg.1 | ENST00000442633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014092.hg.1 | ENST00000442633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014092.hg.1 | ENST00000442633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014092.hg.1 | ENST00000442633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014092.hg.1 | ENST00000442633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014092.hg.1 | ENST00000442633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014092.hg.1 | NR_037950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014092.hg.1 | NR_037950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014092.hg.1 | NR_037950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014092.hg.1 | NR_037950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014092.hg.1 | NR_037950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014092.hg.1 | NR_037950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014092.hg.1 | NR_037950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | ENST00000273695.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | ENST00000273695.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | ENST00000273695.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | ENST00000273695.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | ENST00000273695.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | ENST00000273695.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | ENST00000273695.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | ENST00000273695.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | ENST00000273695.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | ENST00000273695.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | ENST00000440822.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | ENST00000440822.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | ENST00000440822.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | ENST00000440822.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | ENST00000440822.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | ENST00000446879.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | ENST00000446879.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | ENST00000446879.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | ENST00000446879.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | ENST00000446879.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | ENST00000446879.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | ENST00000446879.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | ENST00000446879.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | ENST00000446879.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | ENST00000446879.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | ENST00000454715.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | ENST00000454715.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | ENST00000454715.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | ENST00000454715.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | ENST00000454715.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | ENST00000454715.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | ENST00000454715.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | ENST00000454715.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | ENST00000454715.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | ENST00000454715.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | ENST00000454715.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | NM_001204897.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | NM_001204897.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | NM_001204897.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | NM_001204897.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | NM_001204897.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | NM_001204897.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | NM_001204897.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | NM_001204897.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | NM_001204897.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | NM_001204897.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | NM_001204897.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | NM_001204898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | NM_001204898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | NM_001204898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | NM_001204898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | NM_001204898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | NM_001204898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | NM_001204898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | NM_001204898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | NM_001204898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | NM_001204898.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | NM_138461.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | NM_138461.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | NM_138461.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | NM_138461.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | NM_138461.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | NM_138461.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | NM_138461.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | NM_138461.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | NM_138461.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | NM_138461.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014095.hg.1 | NM_138461.3 | Refseq |

TM4SF19
Genomic Map