Assay Details
| Gene Symbol: GIT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135214.1 | NP_001128686.1 | 14 - 15 | 1559 | 74 | |
| NM_057169.3 | NP_476510.1 | 14 - 15 | 1559 | 74 | ||
| XM_005253997.3 | XP_005254054.1 | 14 - 15 | 1551 | 74 | ||
| XM_017020259.1 | XP_016875748.1 | 14 - 15 | 1551 | 74 | ||
| XM_017020260.1 | XP_016875749.1 | 14 - 15 | 1551 | 74 | ||
| GenBank mRNA | AF124491.1 | - | 14 - 15 | 1394 | 74 | |
| AK303198.1 | - | 13 - 14 | 1260 | 74 | ||
| AK308304.1 | - | 14 - 15 | 73 | |||
| CR749340.1 | - | 1 - 2 | 140 | 74 | ||
More Information
Set Membership: |
|

GIT2
Genomic Map